GeneChip™ HT MG-430 PM Array Plate
Applied Biosystems™

GeneChip™ HT MG-430 PM Array Plate

Proven performance from the industry standard The GeneChip™HT MG-430 PM 96-Array Plate enables high-throughput expression profiling of multiple samples usingRead more
Have Questions?
Catalog NumberQuantity
9012581 x 96-array plate
Catalog number 901258
Price (INR)
-
Quantity:
1 x 96-array plate
Proven performance from the industry standard
The GeneChip™HT MG-430 PM 96-Array Plate enables high-throughput expression profiling of multiple samples using the same industry-standard content as the GeneChip Mouse Genome 430 2.0 Array.*

The HT MG-430 PM Array Plate enables you to:
• Measure gene expression of more than 39,000 transcripts on a single array
• Get accurate and reproducible data by using multiple independent measurements for each transcript

Key Benefits
• Enables increased productivity and efficiency through parallel processing
   - Process 16, 24, or 96 samples on a single array plate
• Same industry-leading performance and content as the Mouse Genome 430 2.0 Array in cartridge format
   - Strong signal and fold change correlation to previous designs
• Multiple independent measurements per transcript for increased confidence in your results
   - Interrogates more sequence than a single probe measurement
• Conveniently packaged
   - Individual 16-, 24-, and 96-array plates available
   - All processing trays included

Content Profile
Sequences used in the design of the array were selected from GenBank™ dbEST and RefSeq. The sequence clusters were created from the UniGene database (Build 107, June 2002) and refined by analysis and comparison with the publicly available draft assembly of the mouse genome from the Whitehead Institute Center for Genome Research (MGSC, April 2002).

*The HT MG-430 PM Array Plate only retains perfect match (PM) probes from the cartridge design. Mismatch (MM) probes were removed.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity1 x 96-array plate
TypeHT MG-430 PM Array Plate
ArrayTranscriptome Profiling
Format96-array Plate
Number of Arrays96 arrays
SpeciesMouse
Unit SizeEach

Frequently asked questions (FAQs)

Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?

The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.