Axiom™ Genome-Wide CHB 2 Array Plate
Applied Biosystems™

Axiom™ Genome-Wide CHB 2 Array Plate

Axiom Genome-Wide Population-Optimized Human Arrays are genotyping panels that offer the best genetic coverage of rare and common variants forRead more
Have Questions?
Catalog NumberQuantity
9018421 plate
Catalog number 901842
Price (INR)
1,221,856.00
Each
Quantity:
1 plate
Price (INR)
1,221,856.00
Each
Axiom Genome-Wide Population-Optimized Human Arrays are genotyping panels that offer the best genetic coverage of rare and common variants for genome-wide disease association studies.

The Axiom Genome-Wide CHB 2 Array Plate is designed to target rare variants (MAF 2-5%) as a complement to Axiom Genome-Wide CHB 1 Array Plate. The Axiom Genome-Wide CHB 1 and CHB 2 Array Set Bundle enables the most comprehensive commercially available genome-wide coverage in CHB populations.

Benefits of Axiom Genome-Wide Population-Optimized Arrays
Each array plate was designed to achieve ˜90% genomic coverage (r2 >0.8) in each target population, while covering markers with MAF of 1-5% in areas such as:
• Critical biological categories (e.g., coding SNPs)
• Biological processes (e.g., drug metabolizing genes)
• Disease categories (e.g., cardiovascular disease, cancer, immunity/inflammation, MHC, and CNS)
• Common and rare SNPs and insertions/deletions (indels) from the International HapMap Project, 1000 Genomes Project, and published disease associations
• Genotype-tested genomic content proven to give informative and reliable results
• Markers preselected for coverage in critical biological categories, such as coding SNPs, and biological pathways, such as drug metabolizing or cardiovascular genes
• High sample pass rate, call rate, and reproducibility
• Compatible with the Axiom 2.0 Reagent Kit, GeneTitan™ MC Instrument, automated or manual workflow, and Genotyping Console™ Software

Other Maximum Genome-Wide Coverage Arrays
These array plates are designed to maximize whole-genome coverage of common and rare alleles, including SNPs within 10 kb of genes.
• Axiom Genome-Wide CEU 1 Array Plate - The first cost-effective array to maximize coverage of rare variants (MAF >1%) for European populations
• Axiom Genome-Wide ASI 1 Array Plate - The first array designed to maximize genomic coverage of rare alleles (MAF >1%) of a consensus East Asian (JPT+CHB) genome
• The Axiom Genome-Wide CHB 1 and CHB 2 Array Set Bundle - This set bundle contains the Axiom Genome-Wide CHB 1 Array Plate and the Axiom Genome-Wide CHB 2 Array Plate. The Axiom Genome-Wide CHB 1 Array Plate maximizes genomic coverage of common alleles (MAF >5%) of the Han Chinese genome. The Axiom Genome-Wide CHB 2 Array Plate is designed to target rare variants (MAF 2-5%) as a complement to Axiom Genome-Wide CHB 1 Array Plate. The Axiom Genome-Wide CHB 1 Array Plate designed to target common variants (MAF >5%) can be accessed at 901764.
• Axiom Genome-Wide PanAFR Array Plate Set - The first array to offer pan-African genomic coverage, with ≥90% genetic coverage of common and rare variants (MAF >2%) of the Yoruba (West African) genome and >85% coverage of common and rare variants (MAF >2%) of the Luhya and Maasi (East African) genomes. This array set also offers high genomic coverage (>85%) in admixed populations with West African ancestry.

Please visit our NetAffx Analysis Center for more information on microarray analysis

Don't see what you're looking for? Learn how to create an Axiom myDesign™ Array Plate tailored for your study.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?

The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.