Axiom™ Genome-Wide AFR 1 Array Plate
Applied Biosystems™

Axiom™ Genome-Wide AFR 1 Array Plate

We have got you coveredAxiom World Arrays are designed for genome-wide imputation analysis with a focus in disease-associated genes. TheRead more
Have Questions?
Catalog NumberQuantity
9018481 plate
Catalog number 901848
Price (INR)
-
Quantity:
1 plate
We have got you covered
Axiom World Arrays are designed for genome-wide imputation analysis with a focus in disease-associated genes. The arrays include additional markers that are specific to multiple HapMap populations. These arrays have the industry's highest coverage of rare variants to replicate studies of specific loci and/or genes that have been identified in previous genome-wide association studies. The use of imputation enables a highly cost-effective, budget conscious alternative to other commercially available arrays.

The Axiom Genome-Wide AFR 1 Array Plate is designed for maximum coverage of Admixed population and diseases of West African and European ancestry using intelligent pair-wise and imputation design tools.

Benefits of Axiom World Arrays:
• Saturated coverage in over 5,000 gene regions previously identified as disease-associated: SNPs selected from disease and drug response GWAS databases including the National Human Genome Research Institute (NHGRI) Catalog of Published Associations, the Human Genome Epidemiology Navigator (HuGE), the Pharmacogenetics Knowledge Base (PharmaGKB), and the Pharmacogenetics Membrane Transporter (PMT) database
• Common and rare (MAF ∼1%) SNPs and indels from the International HapMap Project and 1000 Genomes Project
• Intelligent, innovative SNP selection maximizes efficient imputation of millions of additional SNPs
• GWAS, replication and fine mapping in one experiment to save time and cost
• Genotype-tested genomic content proven to give informative and reliable results
• High sample pass rate, call rate, and reproducibility
• Compatible with the Axiom 2.0 Reagent Kit, GeneTitan™ MC Instrument, automated or manual workflow, and Axiom Analysis Suite

Other Available Arrays
World Array 1: Axiom Genome-Wide EUR 1 Array Plate – European ancestry
World Array 2: Axiom Genome-Wide EAS 1 Array Plate – East Asian ancestry
World Array 4: Axiom Genome-Wide LAT 1 Array Plate – Admixed population of Native American, European, and West African ancestry
Don't see what you're looking for? Learn how to create an Axiom myDesign™ Array Plate tailored for your study.

Required Products
Axiom Analysis Suite
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity1 plate
ArrayGenotyping
Format96-array Plate
Number of Arrays96 arrays
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

Is the hybridization mix for cartridges the same as the hybridization mix for PEG arrays (array plates)?

The hybridization mix for PEG arrays is different from the hybridization mix for cartridges. The concentrations of the hybridization mix are different and in addition, DMSO is not added into the hybridization mix for PEG arrays because it can affect the glue that holds the PEG to the plate.

What is the starting amount of DNA required for Axiom arrays?

All Axiom arrays (except for Axiom Genome-Wide Pan-African Array Set) require a total of 200 ng DNA. The Axiom Genome-Wide Pan-African Array Set requires a total of 300 ng DNA, or 100 ng DNA per array (there are 3 arrays).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What are the sample requirements for Axiom array processing at the Thermo Fisher Scientific Microarray Research Services Laboratory (MRSL)?

For information regarding our in-house MRSL requirements, please contact us via phone at 888-362-2447 (in North America) or via email at BioinformaticsServices@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.