GeneChip™ Arabidopsis Gene 1.0 ST Array
Applied Biosystems™

GeneChip™ Arabidopsis Gene 1.0 ST Array

The GeneChip™ Arabidopsis Gene 1.0 ST Arrays enable you to:• Measure expression, across the entire gene, with higher resolution andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
9019166 arrays
90191530 arrays
Catalog number 901916
Price (INR)
-
Number of Arrays:
6 arrays
The GeneChip™ Arabidopsis Gene 1.0 ST Arrays enable you to:
• Measure expression, across the entire gene, with higher resolution and accuracy than with classical 3'-biased microarray solutions
• Get accurate and reproducible data by using multiple independent measurements for each transcript

Background
Model and applied research organisms are valuable for comparative genomics research, evolutionary biology, and continue to play a critical role in deciphering the molecular mechanisms underlying human disease, and agricultural crop improvement. The GeneChip Gene 1.0 ST Arrays have been developed for the analysis of a wide range of model and applied research organisms. These organisms are the latest additions to the growing family of Gene Expression Microarrays offering whole-transcript coverage. The GeneChip Gene 1.0 ST Arrays were designed in collaboration with influential researchers, such as Alan Archibald, Head of the Division of Genetics and Genomics at the Roslin Institute (Porcine Array design), Leonard Zon, Director of Stem Cell Research Program, and Yi Zhou, Genomic Core Director of the Stem Cell Research program at Children's Hospital Boston and Harvard Medical School in Boston (Zebrafish Array design).

Key benefits
Highest transcript coverage – get confident expression measurements of well-annotated content with up to 26 probes per transcript
Whole-transcriptome analysis – capture the transcript isoforms you may miss with 3'-biased expression designs
High data correlation – achieve high inter- and intra-array strip signal correlation (R >0.99)

Proven performance from the industry standard
GeneChip Gene 1.0 ST Arrays offer whole-transcriptome coverage for selected model and applied research organisms. All designs are based on the most recent genomic content and offer the highest probe coverage (up to 26 probes across the full length of the gene). This allows for accurate detection for whole-transcriptome microarray analysis and provides higher resolution and accuracy than other classical 3'-biased microarray solutions on the market. The whole-transcriptome analysis approach enables researchers to detect multiple transcript isoforms, including those that might be missed using a 3'-biased expression design, such as splice variants, non-polyadenylated transcripts, transcripts with alternative polyadenylation sites, and truncated transcripts.

Complete Reagents, Software and Instrumentation Solution for Optimal Array Performance
For convenience and complete support, the Gene 1.0 ST Array is provided as part of a comprehensive solution that includes GeneChip reagents and instrumentation:
• GeneChip WT Sense Target Labeling and Control Reagents. For detailed information on the procedure of the assay and performance information, please refer to the WT Sense Target Labeling Assay Manual
• GeneChip Fluidics Station 450 for complete walk-away array processing to obtain the highest levels of reproducibility
• GeneChip Scanner 3000 7G with the optional Autoloader for array image acquisition

We offer several tools to help you in your data analysis, including GeneChip Expression Console; software, the NetAffx Analysis Center, and the Integrated Genome Browser (IGB).

Expression Console Software is an easy-to-use application that enables probe set summarization as well as preliminary data quality evaluation. A simple workflow enables the user to quickly analyze the data (see figure). The resulting data can be further analyzed using software applications from GeneChip-compatible™ software providers.

The NetAffx Analysis Center, with regularly updated biological and functional annotations of probe sets, is the most comprehensive resource for array annotations and probe sequence information. Flexible query tools and external links allow researchers to drill down into genes and annotations of interest. This resource makes it easy to interpret microarray results and quickly design downstream studies.

Workflow
Researchers can also use IGB to visualize results in a genomic context. Genomic annotations including RefSeq sequences, SNPs and genomic locations from various sources can be viewed alongside microarray gene expression signal data.

The Gene 1.0 ST Array data analysis workflow is similar to the workflow for 3' gene expression analysis, utilizing Expression Console 1.1, third-party GeneChip-compatible software and annotation tools in the NetAffx Analysis Center and IGB.

Content profile
GeneChip Gene 1.0 ST Arrays provide the latest coverage of the transcribed genome. We use a comprehensive collection of information sources to design probes that interrogate up to 26 unique sequences of each transcript. Together these 26 unique 25-mer probes interrogate up to 650 bases per transcript. This high probe coverage across the entire transcript results in superior performance and data confidence as well as the ability to update your experimental data as the understanding of each genome and transcriptome grows.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineGeneChip
Quantity6 arrays
TypeArabidopsis Gene 1.0 ST Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays6 arrays
SpeciesArabidopsis
Unit SizeEach

Frequently asked questions (FAQs)

Are pseudogene databases included in the design of expression arrays?

Pseudogene databases were not included in the design of expression arrays.

How long can I store labeled cDNA when working with expression microarrays?

Labeled material can be stored for 2 weeks at -20 degrees C.

Is the protocol same for Exon 1.0 St and Gene 1.0 St arrays?

The protocols for the Gene 1.0 ST and Exon 1.0 ST arrays are the same until the fragmentation and labeling steps.
Depending on the input amount, please refer to the GeneChip WT Pico Reagent Kit or the GeneChip WT PLUS Reagent Kit for the protocol:
- WT Pico: 50 - 500ng of total RNA
- WT PLUS: ≥ 100pg of total RNA (approx. 10 cells)
From the hybridization step onwards, the differences in protocol are due to the format of the arrays. The Exon array is a 49 format array, whereas the Gene 1.0 ST array is a 169 format array.
For the fluidics step the following scripts should be used depending upon which array you are using:
- Gene 1.0 St array: FS450_0001
- Exon 1.0 St array: FS450_0007

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I install library files for my Gene 2.0 ST Array?

To install library files for the Gene 2.0 ST Array for Command Console, simply run the executable library file installer.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

How do I install library files for Gene 1.0 ST Array?

Installing the library files for the Gene 1.0 ST Array for AGCC requires the use of the Affymetrix Library File Importer which converts GCOS library files into AGCC format, then imports the files to the Command Console library file folder. The Library File Importer can convert files from the local GCOS installation, from a GCOS server on a remote Windows Server, or from a GCOS library file package on a CD or disk drive.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.