Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Actual product may vary
Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Automation Tips for Tecan™ Liquid Handling Platforms
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Have Questions?
Catalog NumberVolume (Metric)SterilityTip StyleQuantity
902-25150 μLSterileConductive96 Tips/Insert, 10 Inserts/Pk.
Catalog number 902-251
HSN 84799090
Price (INR)
22,532.00
Case
Add to cart
Volume (Metric):
50 μL
Sterility:
Sterile
Tip Style:
Conductive
Quantity:
96 Tips/Insert, 10 Inserts/Pk.
Price (INR)
22,532.00
Case
Add to cart
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
Features:
  • Available as sterile, filtered and/or liquid sensing.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
Volume (Metric)50 μL
FilteredNon-filtered
SterilitySterile
ColorBlack
Rack ColorPurple
AutoclavableNot Autoclavable
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
FormatReload Deck
Tip StyleConductive
PackagingBlister Pack
Quantity96 Tips/Insert, 10 Inserts/Pk.
For Use With (Equipment)Tecan Systems with LiHa Head
Fit for PurposeSterile
Unit SizeCase