GeneChip™ Human Gene 2.0 ST Array
Applied Biosystems™

GeneChip™ Human Gene 2.0 ST Array

The GeneChip™ Human Gene 2.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and longRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
9021126 arrays
9024992 arrays
90211330 arrays
Catalog number 902112
Price (INR)
-
Number of Arrays:
6 arrays
The GeneChip™ Human Gene 2.0 ST Array provides the most accurate, sensitive, and comprehensive measurement of protein coding and long intergenic non-coding RNA transcripts.

Comprehensive design
Keeping pace with the research community's understanding of the transcriptome, we have designed whole-transcript arrays that include probes to measure both messenger (mRNA) and long intergenic non-coding RNA transcripts (lincRNA). These whole-transcript array designs provide a complete expression profile of mRNA as well as the intermediary lincRNA transcripts that impact the mRNA expression profile.

Research over the past 20 years has predominantly focused on protein coding messenger RNA transcripts and their role in cellular processes, such as disease and development. Recently researchers have identified more that 10,000 transcripts (>200 bases) with little or no protein coding potential. Only a small fraction of these non-coding RNAs has functional annotations to date. However, there is ample evidence that differential expression of lincRNAs plays an important role in the genesis and progression of disease and that aberrant expression of these molecules have also been linked to cancer. Recent advancements in transcriptome profiling provided evidence of the association of lincRNAs in a diverse range of cellular functions:
• Regulation of mRNA transcription
• Regulation of mRNA post-transcriptional modifications
• Occlusion/recruitment of transcription factor binding
• Activation and transportation of transcription factors
• Interaction with accessory proteins
• Guide protein complexes to locations in the genome

Key benefits
• Comprehensive coverage provides the best opportunity to discover interesting biology
• >30,000 coding transcripts
• >11,000 long intergenic non-coding transcripts
• Reproducible: Intra-lot correlation coefficient =0.99

Content profile
Since the design of the Human Gene 1.0 ST Array, there have been a massive number of new lincRNA that have been identified by the research community. In order to provide the research community with a tool that can measure the differential expression of this exciting class of RNA transcripts, we have designed the Human Gene 2.0 ST Array. To supplement the lincRNA data contained in RefSeq, we used sequences and transcripts from lncRNA db www.lncrnadb.com and Broad Institute, Human Body Map lincRNAs and TUCP (transcripts of uncertain coding potential catalog) lincRNAs
For Research Use Only. Not for use in diagnostic procedures.
Specifications
IncludesProbe
Product LineGeneChip
Quantity6 arrays
TypeHuman Gene 2.0 ST Array
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays6 arrays
SpeciesHuman
Unit SizeEach

Frequently asked questions (FAQs)

What is contained in the tab-delimited format of the GeneChip probe sequence download file?

The tab-delimited probe sequence file contains the following information:
-Probe Set Name
-Probe X: The X coordinate of the probe sequence on the GeneChip probe array.
-Probe Y: The Y coordinate of the probe sequence on the GeneChip probe array.
-Probe Interrogation Position: The base position on the consensus/exemplar sequence where the central base of the probe aligns, which is the 13th base of a 25mer probe.
-Probe Sequence: The 25-base perfect match sequence.
-Target Strandedness: The sense/antisense orientation of the target sequence that can hybridize with the probe sequence.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What is the NetAffx Analysis Center?

The NetAffx Analysis Center used to contain information and files regarding array content, probe sets, and functional annotations. However, the NetAffx Analysis Center has been retired. Much of the content provided by the NetAffx Analysis Center can now be accessed on the arrays’ thermofisher.com product pages or by contacting Technical Support (techsupport@thermofisher.com).

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What reagent kit should I use with my array?

Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

Are pseudogene databases included in the design of expression arrays?

Pseudogene databases were not included in the design of expression arrays.

How long can I store labeled cDNA when working with expression microarrays?

Labeled material can be stored for 2 weeks at -20 degrees C.