Product Image
Applied Biosystems™
PrimeView Human Gene Expression Assay
The PrimeView Human Gene Expression Assay enables you to: • Measure gene expression of more than 36,000 transcripts and variantsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90248730 arrays
90248610 arrays
Catalog number 902487
Price (INR)
-
Number of Arrays:
30 arrays
The PrimeView Human Gene Expression Assay enables you to:
• Measure gene expression of more than 36,000 transcripts and variants per sample
• Get accurate and reproducible gene expression data by using multiple independent measurements for each transcript
Proven performance from the industry standard
The PrimeView Human Gene Expression Assay enables expression profiling using probe sets with an emphasis on established, well annotated content. Sequences used in the design of the assay were selected from the RefSeq version 36, UniGene database 219, and full-length human mRNAs from GenBank™.
Key benefits
• Offers excellent gene expression specificity and reproducibility
• Produces reliable results with multiple independent measurements per transcript
• Provides complete coverage of the annotated genome
• Highest transcript coverage in the industry with the most recent annotation
Content profile
• The PrimeView Human Gene Expression Assay is comprised of more than 530,000 probes covering more than 36,000 transcripts and variants, which represent more than 20,000 genes mapped through RefSeq or via UniGene annotation.
• The EST and mRNA sequences used in the design were clustered and assembled to create consensus sequences that represent alternative splice forms. Each assembly was then analyzed for orientation and alternative 3' end evidence.
• Content was chosen to cover all well-annotated genes and transcripts from RefSeq v36 and by leveraging all available EST and mRNA evidence that fall into the same clusters, to rigorously detect alternate 3' ends of those well-annotated genes.
• More than 1,000 probe sets represent transcripts that have no official gene symbol in UniGene, but are based on predicted RefSeq sequences and UniGene clusters with good evidence of actual transcription.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Quantity30 reactions
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays30 arrays
SpeciesHuman
Unit SizeEach
Frequently asked questions (FAQs)
What reagent kit should I use with my array?
Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.