Product Image
Applied Biosystems™
Clariom™ D Assay, mouse
Clariom™ D assays, mouse were previously known as GeneChip™ Mouse Transcriptome Array 1.0 (MTA 1.0.)Accelerate your biomarker discovery from deepRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90251310 arrays
90251430 arrays
Catalog number 902513
Price (INR)
-
Number of Arrays:
10 arrays
Clariom™ D assays, mouse were previously known as GeneChip™ Mouse Transcriptome Array 1.0 (MTA 1.0.)
Accelerate your biomarker discovery from deep within the transcriptome with Clariom D assays, the next-generation of transcriptome-level expression profiling tools. Clariom D assays provide a highly detailed view of the transcriptome and offer the fastest path to actionable results. Available for human, mouse, and rat, Clariom D assays allow translational research scientists to generate high-fidelity biomarker signatures quickly and easily. Based on industry-leading microarray technology, the novel Clariom D assay design provides the most intricate transcriptome-wide gene- and exon-level expression profiles, including the ability to detect alternative splicing events of coding and long non-coding (lnc)RNA, in a single three-day experiment.
Expand your potential to discover novel, informative biomarkers.
The number of known transcribed genes has expanded rapidly in recent years providing more sources for actionable biomarkers, such as transcript variants and lncRNA, that can be used for clinical utility and advancing our understanding of disease mechanisms. Such biomarkers can be missed by lengthy, complex, and costly sequencing and targeted expression approaches, leading to irreproducible signatures and wasted time and money.
With full coverage of the transcribed genome including all known coding and non-coding splice variants, compatibility with clinical sample types, and flexible data analysis software, Clariom D assays are the premier tools for translational researchers performing complex expression biomarker discovery studies and wanting the fastest path to robust, clinically relevant, and actionable results.
Get all the data you need.
• Rapidly identify complex disease signatures using >214,000 transcripts sourced from the largest number of public databases, the most comprehensive coverage of the mouse transcriptome.
• Confidently detect genes, exons, and alternative splicing events that give rise to coding RNA and lncRNA isoforms.
• Detect rare and low-expressing transcripts otherwise not detected by common sequencing approaches.
• Go from data to insight in minutes with intuitive, highly visual, free analysis software.
When you have precious samples, get it right the first time.
• Utilize RNA from various sample types including blood, cells, and fresh/fresh-frozen tissues.
• Preserve sample integrity and reduce data variability with an assay that does not require a globin or rRNA removal step.
Clariom D solutions are available in a single sample (cartridge array) format for use on the GeneChip™ 3000 instrument system and include reagents and fast, simple Transcriptome Analysis Console (TAC) software to analyze and visualize global expression patterns of genes, exons, pathways, and alternative splicing events.
Get the coverage you require, the reproducibility you need, and the insights you want to act on your discoveries.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineApplied Biosystems
Quantity10 reactions
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
TypeD Assay
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays10 arrays
SpeciesMouse
Unit SizeEach
Contents & Storage
• Poly-A RNA Control, store at -20°C
• Hybridization Control Kit, store at -20°C
• WT Terminal Labeling Kit, store at -20°C
• WT Amplification Kit, Module 1, store at -20°C
• WT Amplification Kit, Module 2, store at 4°C
• Clariom D 10 Arrays, mouse, store at 4°C
Frequently asked questions (FAQs)
What reagent kit should I use with my array?
Please refer to the Microarray Reagent Guide for Arrays and Expression Kits to match the correct reagents your array.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why is it important to use Tough-Spots® labels when using GeneChip cartridge arrays?
Tough-Spots® labels are small adhesive stickers used to temporarily seal the backs of cartridge arrays during the overnight hybridization step. They are required to prevent loss of volume due to evaporation through the septa. We recommend using Tough-Spots® labels on Rolls from USA Scientific (Item No. 9185-0000)
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What are the respective hybridization volumes based on array format for Affymetrix gene expression arrays?
Proper hybridization volume is critical to obtaining an even signal across a given array. Too little volume can lead to black circles in the middle of the array. Too much volume can leak out of the back of the array. The correct hybridization volume leaves enough room for a small air bubble to circulate around the array surface during the overnight hybridization. Here are the recommended hybridization and fill volumes based on the array format:
Array Format; Hybridization Volume; Fill Volume
- 49 Format (Standard); 200 µL; 250 µL
- 64 Format; 200 µL; 250 µL
- 100 Format (Midi); 130 µL; 160 µL
- 169 Format (Mini); 80 µL; 100 µL
- 400 Format (Micro); 80 µL; 100µL
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
Why is strand-specificity important when performing Clariom D and Clariom S assays?
Clariom D arrays have probes that cover all known regions of transcription including probes in overlapping regions from both strands. To obtain strand-specific information from the Clariom D arrays, the WT Pico and WT Plus reagents (which are strand-specific) must be used. This is important because without strand-specific reagent, it would not be possible to decipher the source strand of DNA, which makes it challenging to untangle true gene- and exon- level expression and alternative splicing events.
Strand-specificity is significantly less important for customers interested in gene-level only information (i.e., those using Clariom S) as compared to customers who want to understand the complexities of the whole transcriptome including identifying antisense transcripts and ncRNA (i.e., those using Clariom D). While strand-specificity is less important for gene-level expression only, probes within regions of overlapping transcription from both strands are avoided in the Clariom S array design (unlike Clariom D). This is important because if Clariom S did not preserve strand-specificity, there could be an overestimation of gene-level expression causing false positive or negative results. With Clariom S having a "stranded" design, it does not necessarily need a strand-specific reagent kit.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.
What is an Event Score in TAC 4.0 Software?
TAC 4.0 includes two algorithms for identifying alternative splicing events: the TAC 2.0 algorithm and the new EventPointer. Algorithmic determination of alternate splicing remains a challenging problem. TAC 4.0 supports two different approaches that have different sets of strengths and weaknesses. After considerable testing, the new TAC 4.0 “'Event Score” leverages both previous TAC 2.0 event estimation score and Event Pointer p-value and sorts the most likely alternative splicing events to the top. Of course, the TAC 2.0 event score and EventPointer p-values remain individually available.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.