GeneChip™ Medicago Transcriptome Pico Assay
GeneChip™ Medicago Transcriptome Pico Assay
Applied Biosystems™
GeneChip™ Medicago Transcriptome Pico Assay
GeneChip™ Medicago Transcriptome Pico Assay, which utilizes 100-2,000 pg of input RNA) is designed for agriculture researchers studying legume genomicsRead more
Have Questions?
Catalog NumberQuantity
90287930 samples
Catalog number 902879
Price (INR)
980,209.00
Each
Add to cart
Quantity:
30 samples
Price (INR)
980,209.00
Each
Add to cart
GeneChip™ Medicago Transcriptome Pico Assay, which utilizes 100-2,000 pg of input RNA) is designed for agriculture researchers studying legume genomics and symbiotic relationships between nitrogen-fixing bacteria and plants. Coupled with our free, intuitive Transcriptome Analysis Console (TAC) Software, it is the fastest and most effective whole-transcriptome profiling approach for legume researchers who want results within a crop season.
• In a single experiment, measure expression changes of M. truncatula reference cultivars A17 and R108, and M. sativa (alfalfa).
• With the right results the first time, have confidence in low-level expression data with higher reproducibility than RNA-Seq.
• Within minutes, analyze and visually interpret the expression of genes, exons, and transcript isoforms.
For additional information, see the GeneChip Medicago transcriptome solution data sheet.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Product LineGeneChip
Quantity30 samples
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays30 arrays
SpeciesMedicago
Unit SizeEach
Frequently asked questions (FAQs)
Is there a recommended RIN (RNA integrity number), based on Bioanalyzer, for RNA quality for IVT and WT assays?
For IVT assays, the RIN should be 6 or higher. For WT assays, there are no recommendations based on assay priming.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.