GeneChip™ Medicago Transcriptome Assay
Applied Biosystems™
GeneChip™ Medicago Transcriptome Assay
GeneChip™ Medicago Transcriptome Pico Assay, which utilizes 100-2,000 pg of input RNA) is designed for agriculture researchers studying legume genomicsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNumber of Arrays
90288010 arrays
90288130 arrays
Catalog number 902880
Price (INR)
381,521.00
Each
Number of Arrays:
10 arrays
Price (INR)
381,521.00
Each
GeneChip™ Medicago Transcriptome Pico Assay, which utilizes 100-2,000 pg of input RNA) is designed for agriculture researchers studying legume genomics and symbiotic relationships between nitrogen-fixing bacteria and plants. Coupled with our free, intuitive Transcriptome Analysis Console (TAC) Software, it is the fastest and most effective whole-transcriptome profiling approach for legume researchers who want results within a crop season.
• In a single experiment, measure expression changes of M. truncatula reference cultivars A17 and R108, and M. sativa (alfalfa).
• With the right results the first time, have confidence in low-level expression data with higher reproducibility than RNA-Seq.
• Within minutes, analyze and visually interpret the expression of genes, exons, and transcript isoforms.
For additional information, see the GeneChip Medicago transcriptome solution data sheet.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Microarray Analysis
Product LineGeneChip
Quantity10 samples
ArrayTranscriptome Profiling
FormatArray Cartridge
Number of Arrays10 arrays
SpeciesMedicago
Unit SizeEach