PharmacoScan™ Training Kit, 24-Format
Applied Biosystems™

PharmacoScan™ Training Kit, 24-Format

The PharmacoScan Training Kit includes four 24-format array plates plus reagents, controls, and consumables to process 88 samples and 8 controls.
Have Questions?
Catalog NumberQuantity
903011TS
also known as 903011
88 samples
Catalog number 903011TS
also known as 903011
Price (INR)
2,116,133.00
Each
Quantity:
88 samples
Price (INR)
2,116,133.00
Each

The PharmacoScan Training Kit includes four 24-format array plates plus reagents, controls, and consumables to process 88 samples and 8 controls. Two DNA training plates containing 22 known samples each are also included.

Note: This includes all consumables and reagents already included with the PharmacoScan Assay Kit (903010TS), but adds two training plates containing known DNA controls.

PharmacoScan Solution is an ideal platform for translational researchers in healthcare, pharma, and research organizations looking to understand how genetic variations affect drug absorption, distribution, metabolism, and excretion (ADME).

By pre-emptively screening for risk factors on a microarray, users in research can precisely and reproducibly analyze 4,627 high- to low-evidence markers* within nearly 1,200 genes* in a single assay. The solution includes copy number variation calling and interrogating predictive markers in complex genes. Unlike solutions that analyze only high-evidence markers, PharmacoScan Solution presents a more comprehensive view for genotyping for pharmacogenomic risk.

*Disclaimer: Number of variants genotyped by default as well as variant to gene mapping is refined over time.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)Microarray Analysis
No. of Samples88 samples
Quantity88 samples
TypeTraining Kit
ArrayGenotyping
Format24-array Plate
Number of Arrays24 arrays
SpeciesHuman
Unit SizeEach
Contents & Storage
4 X 24-format array plates. Store at 2-8°C

2 x training plates containing known DNA samples. Store frozen -5 to -30°C.

1 x Reagent Kit (Includes reagents and controls for 4x24-format array plates).Storage can vary for each reagent. Refer to product insert.

Gene Titan instrument consumables for 4x24-format array plates. Store at room temp.

Frequently asked questions (FAQs)

Do you offer an alternative for the discontinued DMET PLUS Assay (Cat. No. 901268)?

We recommend reviewing the Microarrays for Preemptive Pharmacogenomics page to determine a potential alternative.

If you have any questions you can email Tech Support at techsupport@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.