Product Image
Applied Biosystems™

GeneChip™ PrimeView™ Global Gene Expression Profile Assay

The GeneChip PrimeView Global Gene Expression Profile Assay enables expression profiling with an emphasis on established, well-annotated content. More thanRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
90540610 Reactions, 10 Arrays
90540730 Reactions, 30 Arrays
Catalog number 905406
Price (INR)
-
Quantity:
10 Reactions, 10 Arrays

The GeneChip PrimeView Global Gene Expression Profile Assay enables expression profiling with an emphasis on established, well-annotated content. More than 36,000 transcripts and variants are measured per sample. Sequences used in the design of the array were selected from RefSeq version 36, UniGene database 219, and full-length human mRNAs from GenBank™.

Features include:

  • Cost-effective global confirmation of pluripotency marker expression
  • Simple-to-use online analysis tool
  • Published method with over 15,000 samples analyzed
  • For use with GeneChip Scanner 3000

In addition, the PluriTest™ Assay now accepts PrimeView array data from GeneChip Scanner 3000.

About the PluriTest Assay

The PluriTest Assay is a bioinformatics assay that compares the transcriptional profile of a sample to an extensive reference set of >450 cell/tissue types, including 223 hESC lines, 41 iPSC lines, somatic cells, and tissues. The online analysis tool confirms pluripotency marker expression via two separate scores: Pluripotency and Novelty. The Pluripotency Score informs the user how strongly a model-based pluripotency signature is expressed in the samples analyzed. The Novelty Score indicates the general model fit for a given sample.

More information on the PluriTest Assay ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ArrayGenechip probe array; Transcriptome Profiling
FormatArray
No. of Reactions10 Reactions
Product LineGeneChip, PrimeView
Quantity10 Reactions, 10 Arrays
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesHuman
For Use With (Application)Stem Cell Analysis
Unit SizeEach
Contents & Storage
• Arrays, purification beads (Module 2): store at 2–8°C
• Module 1, PolyA kit, hybridization kit: store at -20°C
• Fragmentation buffer (Module 2): store at room temperature

Frequently asked questions (FAQs)

Which kit do I need to purchase to run the PluriTest Assay?

Customers should purchase the GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905406 or 905407). 

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What additional reagents do I need to run the GeneChip PrimeView Global Gene Expression Profile Assay, Cat. No. 905406 or 905407?

The GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905406 or 905407) does not include the hybridization, wash, and stain reagents. Hence, the GeneChip Hybridization, Wash, and Stain Kit (Cat. No. 900720) will have to be purchased separately. Alternatively, the GeneChip PrimeView Global Gene Expression Profile Assay (Cat. No. 905400) can be used; it is a complete kit that includes the GeneChip Hybridization, Wash, and Stain kit. 

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.