Rheodyne™ Sample Loops
Actual product may vary
Rheodyne™ Sample Loops
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)For Use With (Equipment)Diameter (Metric) Inner
9055-026500 μLRheodyne Injectors 9010/97250.76 mm
70205 μLRheodyne Injectors 7010/71250.18 mm
702110 μLRheodyne Injectors 7010/71250.30 mm
702220 μLRheodyne Injectors 7010/71250.30 mm
7026500 μLRheodyne Injectors 7010/71250.76 mm
70295 mLRheodyne Injectors 7010/71251.0 mm
7755-02110 μLRheodyne Injectors 7725/7725i0.30 mm
80205 μLRheodyne Injector Model 81250.20 mm
802220 μLRheodyne Injector Model 81250.25 mm
802350 μLRheodyne Injector Model 81250.30 mm
9055-02220 μLRheodyne Injectors 9725/9725i0.25 mm
9055-02350 μLRheodyne Injectors 9725/9725i0.51 mm
9055-025200 μLRheodyne Injectors 9010/97250.51 mm
9055-0271 mLRheodyne Injectors 9010/97250.76 mm
9055-0295 mLRheodyne Injectors 9010/97250.76 mm
Catalog number 9055-026
HSN 90279090
Price (INR)
16,957.00
Each
Volume (Metric):
500 μL
For Use With (Equipment):
Rheodyne Injectors 9010/9725
Diameter (Metric) Inner:
0.76 mm
Price (INR)
16,957.00
Each
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeSample Loop
DescriptionFor Rheodyne 9010/9725; 500 uL; 0.76 mm ID
For Use With (Equipment)Rheodyne Injectors 9010/9725
Diameter (English) Inner0.030 in.
Diameter (Metric) Inner0.76 mm
Volume (Metric)500 μL
Unit SizeEach