S1 Pipet Filler Accessories
Actual product may vary
Thermo Scientific
S1 Pipet Filler Accessories
Incorporate Thermo Scientific™ S1 Pipet Filler Accessories with S1 Pipet Filler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
9057Pipette Filter, Sterile
4580560Pipette Filter, Non-sterile
9064Nosepiece
9065-11Silicone Pipet Gripper
9066-11Wall Mount Stand
9067Table Mount Stand
9068Universal Power Supply
9069Wing Stand
Catalog number 9057
HSN 84799090
Price (INR)
31,861.00
CS
Type:
Pipette Filter, Sterile
Price (INR)
31,861.00
CS
Complete your S1 Pipet Filler with these accessories and prolong its use.
Features:
  • Wall-mounted holder and table stand offer compact storage between applications. Recharge the pipet filler in both stands by plugging in the charger. Store pipet filler when working in laminar flow cabinets to save space.
  • Wing stand allows user to set the pipet filler down without removing or emptying the serological pipet.
  • Easy to change filter prevents aerosol or liquid cross contamination of the pipet together with autoclavable silicone adapter.
  • 1mL Pipet Support ensures that 1 mL pipets will hold as securely as larger sizes.
Note:
Wing stand not available for use or sale within the United States or for importation into the United States.
Specifications
DescriptionHydrophobic Filters, 0.45µm, Sterile
For Use WithS1 Pipette Filler
TypePipette Filter, Sterile
Unit SizeCS