Automation Tips for Tecan™ Liquid Handling Platforms with Te-MO 96 Multichannel Arm
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Sterility
Filtered
Fit for Purpose
908-262PK
also known as 908-262
200 μL
Non-sterile
Non-filtered
Certified Quality
906-262PK
also known as 906-262
50 μL
Non-sterile
Non-filtered
Certified Quality
2 Options
Catalog number 908-262PK
also known as 908-262
Price (INR)
9,512.00
Pack of 960
Volume (Metric):
200 μL
Sterility:
Non-sterile
Filtered:
Non-filtered
Fit for Purpose:
Certified Quality
Price (INR)
9,512.00
Pack of 960
Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Te-MO, Aquarius and Freedom EVO with Te-MO 96 head.
Features:
Available as sterile and filtered.
Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Lot-tested during production on actual workstation for which they are created.
Low percentage coefficient of variation (%CV).
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius