Orion™ Single Junction Epoxy Bodied pH/ATC Triode™
Orion™ Single Junction Epoxy Bodied pH/ATC Triode™
Orion™ Single Junction Epoxy Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ Single Junction Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) Cable
9107BNBNC1 m
9107BNMDBNC (Waterproof)1.5 m
9107WMMDBNC (Waterproof)3 m
Catalog number 9107BN
Price (INR)
-
Connector Types:
BNC
Length (Metric) Cable:
1 m

Thermo Scientific™ Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes are low-maintenance, gel-filled and come with a built-in ATC. They are engineered with an epoxy body for ruggedness and best suited for general purpose samples or for educational institutes where minimal maintenance is required.

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
 
Solutions required 
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Warranty period
6 months from date of purchase
Specifications
DescriptionpH/ATC Triode
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
For Use With (Application)The Thermo Scientific Orion 9107BN Triode 3-in-1 pH/automatic temperature compensation probe has a low maintenance gel filled electrode with built-in ATC.For field or plant use.
For Use With (Equipment)Orion A/A+ models 210, 230, 250, 410, 420, 520, 525, 710, 720, 920
JunctionWick
Length (English)4.7 in.
Length (Metric)120 mm
Length (Metric) Cable1 m
MaterialEpoxy
Reference TypeAg/AgCl
Temperature (English) Operating32°F to 176°F
Connector TypesBNC
Temperature (Metric) Operating0°C to 80°C
TypepH/ATC Probe, BNC Connector
pH Range0 to 14
Unit SizeEach