Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Actual product may vary
Thermo Scientific™
Sterile Pipette Tips for Beckman™ Liquid Handling Systems
Increase performance through the superior tip straightness and low %CV with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip Style
916-261-0550 μLLow Retention
915-021-0520 μLLow Retention
917-021-05130 μLLow Retention
376150 μLStandard
938-021-0540 μLExtended Length, Low Retention
938-261-0550 μLExtended Length, Low Retention
915-261-0520 μLLow Retention
917-261-05200 μLLow Retention
916-021-0540 μLLow Retention
935-261-0530 μLLow Retention
Catalog number 916-261-05
Price (INR)
-
Request A Quote
Volume (Metric):
50 μL
Tip Style:
Low Retention
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)
Specifications
FilteredFiltered
For Use With (Equipment)Beckman Coulter Liquid Handling Systems
FormatRack
SterilitySterile
Tip StyleLow Retention
Volume (Metric)50 μL
Unit SizeEach