Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Actual product may vary
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredTip StyleFit for Purpose
916-262-05PK
also known as 916-262-05
50 μLNon-filteredLow RetentionCertified Quality
919-021PK
also known as 919-021
130 μLFilteredStandardSterile and PCR Ready
919-262PK
also known as 919-262
250 μLNon-filteredStandardCertified Quality
Catalog number 916-262-05PK
also known as 916-262-05
Price (INR)
7,737.00
Pack of 960
Volume (Metric):
50 μL
Filtered:
Non-filtered
Tip Style:
Low Retention
Fit for Purpose:
Certified Quality
Price (INR)
7,737.00
Pack of 960
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Specifications
Certifications/ComplianceISO 9001 and ISO 13485
Clean ClaimsFree of RNase, DNase, DNA, ATP, and endotoxin
Packaging96 tips/rack, 10 racks/pack, 5 packs/case
Quantity4800 tips
AutoclavableAutoclavable
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96
FormatRack
Product LineMultimek, Multimek AP96
SterilityNon-sterile
Tip StyleLow Retention
Volume (Metric)50 μL
Unit SizePack of 960