Orion™ Single Junction Spear Tip Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ Single Junction Spear Tip Glass Bodied Combination pH Electrode

Pierce soft and semi-solid samples like cheese, meat, or fruit with a refillable spear tip pH electrode.
Have Questions?
Catalog NumberConnector Types
9162BNWPBNC
Catalog number 9162BNWP
Price (INR)
-
Connector Types:
BNC

Thermo Scientific™ Orion™ Spear Tip pH Electrodes provide reliable and accurate measurements of routine samples. Best suited for piercing solid/semi-solid samples and measuring small volume samples.

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
 
Solutions required 
Electrode fill solution
900011     4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
 
Electrode storage solution
910001      pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
910199     Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
                 electrode storage solution, 475ml, and 12 mm electrode storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with
900011     4M KCl with Ag/AgCl filling solution 60mL 1 bottle
 
Warranty period
12 months from date of purchase
Specifications
DescriptionWith BNC waterproof connector
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
For Use With (Equipment)Orion pH Meter
Internal ReferenceAg/AgCl
Length (Metric)110 mm
Precision0.02
Connector TypesBNC
MaterialGlass Body
Temperature (Metric) Operating0°C to 90°C
TypeHalf-Cell Electrode, low resistance, BNC connector
pH Range0 to 14
Unit SizeEach