Search
| Catalog Number | Volume (Metric) | Filtered | Tip Style | Fit for Purpose |
|---|---|---|---|---|
| 919-262 also known as 919-262GPD | 250 μL | Non-filtered | Standard | Certified Quality |
| 912-021 also known as 912-021PD | 8 μL | Filtered | Standard | Sterile and PCR Ready |
| 912-261 | 10 μL | Non-filtered | Standard | Sterile |
| 912-262 | 10 μL | Non-filtered | Standard | Certified Quality |
| 916-021 | 40 μL | Filtered | Standard | Sterile and PCR Ready |
| 916-261 | 50 μL | Non-filtered | Standard | Sterile |
| 916-262 | 50 μL | Non-filtered | Standard | Certified Quality |
| 918-021 also known as 918-021PDX | 20 μL | Filtered | Standard | Sterile and PCR Ready |
| 918-261 | 20 μL | Non-filtered | Standard | Sterile |
| 918-261-05 | 20 μL | Non-filtered | Low Retention | Sterile |
| 918-262 | 20 μL | Non-filtered | Standard | Certified Quality |
| 918-262-05 also known as 918-262-05M | 20 μL | Non-filtered | Low Retention | Certified Quality |
| 919-021 also known as 919-021G | 130 μL | Filtered | Standard | Sterile and PCR Ready |
| 919-021-05 | 130 μL | Filtered | Low Retention | Sterile and PCR Ready |
| 919-261 | 250 μL | Non-filtered | Standard | Sterile |
| 919-261-05 | 250 μL | Non-filtered | Low Retention | Sterile |
| 919-261G | 250 μL | Non-filtered | Wide Bore | Sterile |
| 919-262G | 250 μL | Non-filtered | Wide Bore | Certified Quality |
| 2790 | 175 μL | Filtered | Standard | Sterile |
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.