Search
| Catalog Number | Volume (Metric) | Filtered | Sterility | Fit for Purpose |
|---|---|---|---|---|
| 921-261 also known as 921-261CP | 100 μL | Non-filtered | Sterile | Sterile |
| 920-021 also known as 920-021CP | 50 μL | Filtered | Sterile | Sterile and PCR Ready |
| 920-261 also known as 920-261CP | 50 μL | Non-filtered | Sterile | Sterile |
| 920-262 also known as 920-262CP | 50 μL | Non-filtered | Non-sterile | Certified Quality |
| 921-021 also known as 921-021CP | 80 μL | Filtered | Sterile | Sterile and PCR Ready |
| 921-262 also known as 921-262CP | 100 μL | Non-filtered | Non-sterile | Certified Quality |
| 923-021 also known as 923-021G | 150 μL | Filtered | Sterile | Sterile and PCR Ready |
| 923-261 also known as 923-261GCP | 200 μL | Non-filtered | Sterile | Sterile |
| 923-262 also known as 923-262GCP | 200 μL | Non-filtered | Non-sterile | Certified Quality |
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.