Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
Thermo Scientific™
Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Have Questions?
Catalog NumberSterilityFilteredFit for Purpose
935-262PK
also known as 935-262
Non-sterileNon-filteredCertified Quality
Catalog number 935-262PK
also known as 935-262
Price (INR)
28,492.00
Pack of 3840
Add to cart
Sterility:
Non-sterile
Filtered:
Non-filtered
Fit for Purpose:
Certified Quality
Price (INR)
28,492.00
Pack of 3840
Add to cart
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeCertified Quality
For Use With (Equipment)Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Multimek AP384
FormatRack
Product LineBeckman
SterilityNon-sterile
Tip StyleExtended Length
Volume (Metric)30 μL
Unit SizePack of 3840