Finntip™ Non-Filtered 384 Format Pipette Tips  20 μL
Actual product may vary

Finntip™ Non-Filtered 384 Format Pipette Tips 20 μL

Ensure accurate, precise pipetting. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberSterilityFit for PurposeFormat
9400620Non-sterileCertified QualityBulk
9400610Non-sterileCertified QualityRack
9400613SterileSterile & PCR ReadyRack
Catalog number 9400620
HSN 84799090
Price (INR)
3,860.00
PK
Sterility:
Non-sterile
Fit for Purpose:
Certified Quality
Format:
Bulk
Price (INR)
3,860.00
PK

Accurate, precise pipetting with 384-format volume 20 μL. Finntip™ Finntip 20 Pipette Tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Key Features:

  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked, bulk bag or reload tower
  • Sterile tips are supplied in vacuum sealed, sterilized rack

Proven quality

  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Immediate traceability

  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack and reload packaging options

Note:

Availability may vary by country.

Specifications
Volume (Metric)20 μL
FilteredNon-filtered
SterilityNon-sterile
ColorPurple
ModelFinntip 20
Length (English)1.3 in.
Length (Metric)3.3cm
FormatBulk
Color CodePurple
For Use With (Equipment)Thermo Scientific Finnpipette 20
Product LineFinnTip
Fit for PurposeCertified Quality
Unit SizePK