Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Thermo Scientific™

Orion™ Silver/Sulphide Ion Selective Electrodes

Achieve fast, precise, and cost-effective silver and sulfide ion measurements with dual-electrode technology. Perfect for low-level cyanide and halide titrations.
Have Questions?
Catalog NumberConnector Types
9416BNBNC
Catalog number 9416BN
Price (INR)
-
Connector Types:
BNC

Make quick, simple, accurate and economical measurements of silver and sulfide ions in aqueous solutions with Thermo Scientific™ Orion™ Silver/Sulfide Electrodes. These electrodes have dual electrode-measuring silver and sulfide ions in aqueous solutions, and perform low-level cyanide and halide titrations.

The ThermoFisher Orion Silver/Sulfide electrode complies with SM 4500-S2- G, ASTM D4658-08 for Wastewater measurements.

Features:

  • 9616BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Silver/Sulfide Half-Cell Electrode 9416BN is to be used with the Orion 900200 sure flow double Junction reference electrode.

Solutions required

  • ISA
    • 940011 ISA for silver measurements, 475 mL bottles
    • 941609 SAOB II reagent pack for sulfide measurements, 4 x 475 mL bottles
  • Electrode fill solutions 5 x 60 mL bottles
    • 900062 Optimum Results B for most silver or sulfide measurements and titrations
    • 900061 Optimum Results A for precise sulfide measurements and titrations
    • 900067 Optimum Results C for precise silver measurements and titrations
  • Electrode fill solution for double junction reference half cell electrode
    • 900002 Inner chamber filling solution, 5 x 60 mL bottles
    • 900003 Outer chamber filling solution, 5 x 60 mL bottles
  • Titrant for sulphide titration
    • 948206 0.1 M lead perchlorate solution, 475 mL bottles

Items included with

9619BNWP

948201: Polishing strip. Qty 2.

900062: Optimum Results B, 60 mL 1 bottle

9419BN

948201: Polishing strip. Qty 2.

Warranty period

12 months from date of purchase

Specifications
ApplicationsPotentiometric titrations for chloride and other halides; silver by titration or direct ISE reading.
Certifications/ComplianceUSP 541, SM 4500-Cl D, AOAC 935.47, AOAC 937.09, AOAC 983.14, EN12133, ISO 5943
DescriptionSolid state half-cell with BNC connector
DriftDrift-free Potentials
ElectrodeHalf-cell. Use with Orion Sure-Flow reference 900100 or 900200.
Length (English) Cable3 ft.
Length (Metric) Cable1 m
Connector TypesBNC
For Use With (Equipment)Orion Star Series Meters
JunctionSure-Flow Junction
MaterialEpoxy
Temperature (Metric) Operating0°C to 80°C
TypeHalf-cell Ion Selective Electrode
Unit SizeEach