GFS35(Z) Graphite Furnace and Autosampler for iCE™ 3400 and iCE™ 3500 AA spectrometers
GFS35(Z) Graphite Furnace and Autosampler for iCE™ 3400 and iCE™ 3500 AA spectrometers
Actual product may vary
Thermo Scientific™
GFS35(Z) Graphite Furnace and Autosampler for iCE™ 3400 and iCE™ 3500 AA spectrometers
Offer a pre-aligned graphite furnace and intelligent autosampler to simplify AA analysis and increase productivity with this advanced graphite furnace and autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
942359020052Graphite Furnace/Autosampler
942359020051Rhodium Furnace Upgrade Kit
942359020001Rhodium Furnace/Autosampler
Catalog number 942359020052
Price (INR)
-
Request A Quote
Type:
Graphite Furnace/Autosampler
High performance combined graphite furnace and autosampler module is designed to enable optimum performance when used with Thermo Scientific™ iCE™ 3400 and iCE™ 3500 AA spectrometers. The Thermo Scientific GFS35(Z) Graphite Furnace and Autosampler accessory offers the benefit of a pre-aligned graphite furnace and intelligent autosampler to simplify analysis and increase productivity.
Boost Productivity and Improve Sensitivity
By combining leading edge furnace technology and reliable autosampler functionality, the GFS35 Graphite Furnace and Autosampler module provides our AA users with unparalleled instrument sensitivity and productivity.
Take your analysis even further with the GFS35Z module configuration, with a Zeeman background correction system. Capable of accurately correcting certain types of structured background that the standard Quadline deuterium system cannot adjust for, the GFS35Z module enables maximum flexibility in your AA analysis.
  • Dynamic, optical cuvette temperature control and pre-heated cuvette injection simplifies the process
  • Integrated wash and waste facility eliminates contamination
  • Ash/Atomize self-optimization with Thermo Scientific™ SOLAAR software
  • Maximized sensitivity and increased dynamic working range for Furnace analysis
  • Correction up to 2A of background, with less than 2% residual error even for fast transient signals
  • Configuration to enable Zeeman and Deuterium background correction for maximum flexibility
  • Ideal for analyses characterized by structured background effects, spectral interferences or higher wavelength measurements
The GFS35 module is available for the iCE 3500 AA model only and the GFS35Z module is available for both the iCE 3400 and iCE 3500 AA spectrometers models.
Specifications
For Use With (Equipment)iCE 3400 and 3500 AA Spectrometers
TypeGraphite Furnace/Autosampler
Unit SizeEach