Product Image
Thermo Scientific™
iCE™ 3000 Series Validation Packages
Ensure your Thermo Scientific™ iCE™ 3000 Series AA spectrometer is perfectly calibrated and tested to factory standards with our iCE 3000 Series Validator Packages.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
9423VALR0004Re-calibration CVU Accessory
9423VAL10003FAAS/GFAAS Validator Pack
9423VAL10002GFAAS Validator Pack (Auto OQ)
9423VAL10001FAAS Validator Pack (Auto OQ)
Catalog number 9423VALR0004
Price (INR)
-
Type:
Re-calibration CVU Accessory
Ensure your Thermo Scientific™ iCE™ 3000 Series AA spectrometer is perfectly calibrated and tested to factory standards with iCE 3000 Series Validator Packages, which provide all the necessary materials for complete instrument performance verification.
Easy Path to Instrument Validation and Operation Qualification
  • Validator Logbook provides all the protocols needed to test the system to factory standards and provides a convenient means to store approved test documents
  • Automated calibration unit allows automated Operation Qualification (OQ) of the system
  • Full system verification of iCE 3000 Series AA instruments
  • Kit contains everything needed to complete the verification process
Tailored Kits to Match your Requirements
  • Three validation kits, designed for your specific choice of Thermo Scientific AA instrument, Flame, Furnace or Flame and Furnace
  • iCE 3000 Series Flame AAS Validator Package (FAAS)
  • iCE 3000 Series Graphite Furnace AAS Validator Package (GFAAS)
  • iCE 3000 Series Flame and Graphite Furnace AAS Validator Package (FAAS/GFAAS)
  • Depending on the validation package selected, kit includes: Validator logbook, certified standard solutions and blank, hollow cathode lamps, cuvettes, CD of record templates
Specifications
For Use With (Equipment)iCE 3000 Series Atomic Absorption Spectrometers
TypeRe-calibration CVU Accessory
Unit SizeEach