Labtip Pipette Tips
Actual product may vary
Thermo Scientific™

Labtip Pipette Tips

Basic pipette tips for standard liquid transfer needs.
Have Questions?
Catalog NumberVolume (Metric)FormatPackaging
94300320300 μLBulk1000 tips/bag
Catalog number 94300320
Price (INR)
-
Volume (Metric):
300 μL
Format:
Bulk
Packaging:
1000 tips/bag
Thermo Scientific™ Labtip standard pipette tips are made from 100% virgin polypropylene. These tips are available in 10, 200, 300 and 1000 μL sizes. Available in clear (10 μL and 300 μL), yellow (200 μL), blue (1000 μL), these tips for general lab use across countless liquid transfer applications.
Labtip pipette tips provide:
  • The most common sizes used in a lab: 10, 200, 300 and 1000 μL
  • Non-Filter essential tips for general laboratory usage
  • Non-sterile packaging in both Racked and Bulk options
Packaging Options:
  • Racked, nonsterile - 96 tips/ tray, 10 trays/pack
  • Bulk - 1,000 tips/ bag
  • Bulk - 25,000 tips/bag
Specifications
DescriptionLabtip 300
Length (Metric)5.2cm
Packaging1000 tips/bag
ColorWhite
FilteredNon-filtered
For Use With (Equipment)Thermo Scientific Finnpipette
FormatBulk
MaterialVirgin Polypropylene
Product LineLabTip
SterilityNon-sterile
Volume (Metric)300 μL
Unit SizePack of 1000