Product Image
Thermo Scientific™

Immuno Standard Modules, EB, black

Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Have Questions?
Catalog NumberArray
950294908 x 12
Catalog number 95029490
Price (INR)
-
Array:
8 x 12
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.
Features:
  • Available in eight-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce back scatter light which may be encountered in epifluorescence
  • Transparent or clear format for use in Time Resolved Fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

Specifications
Binding PropertyHydrophilic
CertifiedNo
Detection MethodFluorescence
For Use With (Application)Fluorescence
FormatStrip Plate
No. per Case50
No. per Pack50
Packaging50 Boxes
Plate BlockingNone
Product TypeMicroplate Strip
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Working330 μL
Array8 x 12
ColorBlack
MaterialPolystyrene
No. of Wells96
Well ShapeFlat Bottom (F-well)
Unit SizeCase of 50