Product Image

Reagent Reservoir

Thermo Scientific™ Reagent Reservoirs are designed for use in your pipetting techniques with single and multi-channel pipettes.
Have Questions?
Catalog NumberQuantity
951004710/Pk.
Catalog number 9510047
HSN 39269099
Price (INR)
8,413.00
PK
Quantity:
10/Pk.
Price (INR)
8,413.00
PK

Thermo Scientific™ Reagent Reservoirs are designed to meet all your pipetting needs. We've added features with you in mind: graduation lines guide you on remaining reagent volume, corner pour spouts prevent spilling when emptying the reservoir, and wide stable base helps to prevents tipping.

  • Designed for multiple pipetting applications
  • Trough within a trough design maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
  • Pour-off spouts in all four corners reduce spillage when pouring reagents out of reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
  • 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other
Specifications
Capacity (English)100 mL
Colorwhite
DescriptionReservoir, 100mL, white
Capacity (Metric)100 mL
SterilityNon-sterile
TypeReagent Reservoir
Quantity10/Pk.
AutoclavableNot Autoclavable
DisposableSingle-use
Unit SizePK