Product Image
Thermo Scientific™
Predilution Test Tubes
Prepare a large sample volume for subsequent transfer to microplates using Predilution Test Tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
9520130Predilution Test Tube
9520557Cap Band
Catalog number 9520130
Price (INR)
-
Product Type:
Predilution Test Tube
Prepare a large sample volume for subsequent transfer to microplates with Thermo Scientific™ Predilution Test Tubes.
  • 1.1mL Polypropylene sample tubes
  • Autoclavable for sterile applications
  • Also available: cap bands in strips of eight
Specifications
AutoclavableAutoclavable
Capacity (Metric)1.1 mL
MaterialPolypropylene
Product TypePredilution Test Tube
SterilitySterile
Unit SizeEach