Axiom™ PangenomePro™ Plus SwiftArray™ Kit
Applied Biosystems™
Axiom™ PangenomePro™ Plus SwiftArray™ Kit
PangenomePro delivers comprehensive, population-diverse genome-wide coverage in a high-throughput array format. Designed for inclusive GWAS and imputation studies, PangenomePro helps researchers generate more representative genetic insights across global populations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Samples
9526462x96F
9526441x96F
Catalog number 952646
Price (INR)
-
No. of Samples:
2x96F
The Axiom PangenomePro Plus Array delivers ethnically diverse, genome-wide coverage in a high-throughput 96-array plate format, designed to support whole-genome imputation across global populations and help researchers build more representative genomic datasets. It can serve as a core research tool for human genomics workflows, including GWAS, population health initiatives, polygenic risk scores, pharmacogenomics, and drug discovery.
Features of the Axiom PangenomePro Plus Array include:
  • Marker selection emphasizes broad imputation power across European, African, admixed American, East Asian, and South Asian populations Low input, plug-and-play analysis.
  • Whole-genome analysis from as little as 100 ng genomic DNA with support for target SNP identification, CNV analysis, HLA and blood typing, and 'plug and play' data analysis workflows.
  • GWAS imputation module (~850,000 markers) across ancestral populations
  • Evidence-based clinical/research markers (includes references to public resources such as ACMG 73, ClinVar, NHGRI-GWAS catalog, etc)
  • Pharmacogenomics coverage aligned to guideline and knowledgebase citations (e.g., CPIC, PharmGKB, PharmaADME)
  • Targeted amplification access for challenging/high-homology regions
  • CNV analysis for fixed regions and de novo discovery across the genome
Plus assay advantage
The 'Plus' workflow unlocks pharmacogenomics content and markers for haplotypes in key genes (e.g., CYP2D6) and addresses limitations in other hybridization-based approaches via gene-specific amplification.
Pharmacogenomics Knowledge Base (PharmGKB™)
  • >2,000 markers in Very Important Pharmacogenes as identified by PharmGKB™
  • >300 markers associated with PharmGKB™ level 1A-2B annotations
  • >550 reportable alleles mentioned in Clinical Pharmacogenetics Implementation Consortium (CPIC™) guidelines
  • Targeted amplification to access content in the following genes: CYP1A2, CYP2D6, CYP2B6, CYP2A6, CYP2C19, and those in in highly homologous regions such as CYP2C9, GSTM1, SULT1A1
  • Disease-related variants (Alzheimer's, cancer, cardiovascular disease, cardiometabolic disease, diabetes, neurological disorders)
  • Blood phenotyping variants for blood typing of common and rare blood groups, bleeding disorders, and blood conditions such as sickle cell anemia
  • Human leukocyte antigen (HLA) type of 11 major MHC (major histocompatibility complex) Class I and Class II loci
  • SARS-CoV-2 immune-related markers (human leukocyte antigens, killer immunoglobulin-like receptors, autoimmune markers, and inflammatory markers)
  • Functional variants (loss of function variants, expression quantitative trait loci, nonsynonymous variants, lung phenotypes)
  • Ancestry, wellness, and trait-related variants
Pharmacogenomics Translation Reports
The Pharmacogenomics Translation Reports include:
  • >110 genes
  • >75 genes reporting haplotypes
  • >1000 reportable haplotypes
  • >30 genes with phenotype predictions (e.g., intermediate metabolizer, etc)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Samples2x96F
Quantity192 Reactions
TypeHuman Microarray Genotyping
ArrayGenotyping, DNA, Copy Number
Format96-array Plate
Number of Arrays2 x 96 arrays
SpeciesHuman
Unit SizeEach