Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Thermo Scientific™
Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)FilteredSterilityFit for Purpose
967-261PK
also known as 967-261
1000 μLNon-filteredSterileSterile
965-021PK
also known as 965-021
50 μLFilteredSterileSterile and PCR Ready
Catalog number 967-261PK
also known as 967-261
Price (INR)
20,312.00
Pack of 960
Volume (Metric):
1000 μL
Filtered:
Non-filtered
Sterility:
Sterile
Fit for Purpose:
Sterile
Price (INR)
20,312.00
Pack of 960
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications
ColorClear
FilteredNon-filtered
Fit for PurposeSterile
For Use With (Equipment)Eppendorf epMotion
FormatRack
Product LineEppendorf epMotion
SterilitySterile
Tip StyleAutomation
Volume (Metric)1000 μL
Unit SizePack of 960