NERL™ Reagent Grade Water
Thermo Scientific™

NERL™ Reagent Grade Water

Improve control in critical diagnostic procedures by using this reagent grade water in place of deionized or distilled water.
Have Questions?
Change viewbuttonViewtableView
Quantity:
32 oz.
1 gal.
5 gal.
Catalog number 9800-3
Price (INR)
-
Quantity:
32 oz.
Improve control in critical diagnostic procedures by using Thermo Scientific™ NERL™ Reagent Grade Water in place of deionized or distilled water. This water meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.
  • Ideal for reconstituting chemistry or coagulation products, preparing analytical standards or rinsing delicate electrodes
  • Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration and final filtration to 0.2 micron
  • Essentially free from organic and inorganic particulate and soluble contaminants
  • Contains no preservatives
  • Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water
  • Certificate of Analysis is available online

General Purpose - For in vitro diagnostic or laboratory use only. Not for infusion, injection or irrigation.

For In Vitro Diagnostic Use.  
Specifications
Purity Grade NotesMeets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water. Certificate of Analysis is available online.
Recommended Storage15° to 30°C
Shelf Life12 months
GradeUSP/NF, CLRW
Quantity32 oz.
pH5 to 7
Unit SizeEach