Konelab™ System Drugs of Abuse Test Reagents
Actual product may vary
Konelab™ System Drugs of Abuse Test Reagents
Conduct semiquantitative tests for Drugs of Abuse screening with Thermo Scientific™ Konelab™ System Drugs of Abuse Test Reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
981622Benzodiazepines
981623Cannabinoids
981625Opiates
981680Ethyl Alcohol
981720DoA Neg. Calibrator
981721DoA Calibrator A1
981722DoA Calibrator A2
981723DoA Control Set A
981729DoA Calibrator C1
981730DoA Calibrator C2
981731DoA Calibrator C3
981736DoA Calibration Set E
981737DoA Control Set E
981624Cocaine Metabolite
981728DoA Control Set B
981725DoA Calibrator B2
981726DoA Calibrator B3
981727DoA Calibrator B4
981621Barbiturates
981620Amphetamines
981724DoA Calibrator B1
981683Methadone
Catalog number 981622
Price (INR)
79,437.00
Each
Description:
Benzodiazepines
Price (INR)
79,437.00
Each
Conduct semiquantitative tests for Drugs of Abuse screening with Thermo Scientific™ Konelab™ System Drugs of Abuse Test Reagents. The Konelab™ Drugs of Abuse assays are typically measured from urine. The measurement range is based on calibration; no sample dilution is necessary. For optimal test results, use our system-specified multi-calibrators and controls.
Reagents applied to Konelab analyzers use methods carefully selected according to IFCC and other recommendations. These methods are provided with traceability to international reference materials.
Ordering Information: Not available in the U.S.A.
Drugs of Abuse Test Applications for Konelab™ Analyzers:
  • Amphetamines
  • Barbiturates
  • Benzodiazepines
  • Cannabinoids
  • Cocaine Metabolite
  • Ethyl Alcohol
  • Methadone
  • Opiates
Specifications
DescriptionBenzodiazepines
Detectable AnalytesBenzodiazepines (BZO)
Sample TypeUrine
TypeReagent
Quantity2 x 30 mL
Unit SizeEach