Indiko™ and Konelab™ System D-Dimer to Transferrin  Specific Protein Reagents
Actual product may vary

Indiko™ and Konelab™ System D-Dimer to Transferrin Specific Protein Reagents

Choose from a specific protein test menu of over 20 immunoturbidimetric methods with Thermo Scientific™ Indiko™ and Konelab™ System Specific Protein Reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
981924HbA1c Pretreatment liquid; Kit size: 12 x 20mL
980997SpeciCal Protein Calibrator; Kit size: 5 x 1mL
981250Specitrol; Kit size: 5 x 1mL
981252RF Control; Kit size: 5 x 1mL
981659Hemolyzing Reagent; Kit size: 1000mL
981695HbA1c Control Normal; Kit size: 5 x 1mL
981696HbA1c Control Abnormal; Kit size: 5 x 1mL
981799Specitrol High; Kit size: 5 x 1mL
981915Lipoprotein(a); Kit Size: 1 x 1mL antiserum
981916Lipoprotein(a) Calibrator
981918Lipoprotein(a) Control High; Kit size: 3 x 1mL
981920Rheumatoid factors 2 (RF 2); Kit Size: 2 x 6mL antiserum
981935Haptoglobin; Kit Size: 2 x 3mL antiserum
981936IgA; Kit Size: 2 x 3mL antiserum
981937IgG; Kit Size: 2 x 3mL antiserum
981938IgM; Kit Size: 2 x 3mL antiserum
981940Orosomucoid; Kit Size: 2 x 3mL antiserum
981941Prealbumin; Kit Size: 2 x 3mL antiserum
981943Transferrin; Kit Size: 2 x 3mL antiserum
981917Lipoprotein(a) Control; Kit size: 3 x 1mL
981946D-Dimer, Kit Size: 2 x 8mL
981945sTfR; Kit Size: 2 x 5mL
981871D-Dimer Calibration Set
981868D-Dimer Control
981869D-Dimer Control High
981874Ferritin Calibration Set
981875Ferritin Control
981876Ferritin Control High
981865sTfR Calibration Set
981866sTfR Control
981867sTfR Control High
Catalog number 981924
Price (INR)
15,159.00
Each
Description:
HbA1c Pretreatment liquid; Kit size: 12 x 20mL
Price (INR)
15,159.00
Each

Receive best-in-class methodologies and standardized performance with Thermo Scientific™ Indiko™ and Konelab™ System Specific Protein Reagents, which include a specific protein test menu of 15 immunoturbidimetric methods. All reagents are ready to go and packaged in volume-optimized, barcoded kits; in-house evaluation as well as optimization of reagents guarantees continuous support and optimum results.

All Indiko and Konelab methods are based on carefully-designed protocols for barcoded system reagents. Calibrators and controls are available in separate kits.

Highlights:

  • High-quality reagents are barcoded for automatic identification
  • Most reagents are delivered in ready-to-use liquid form
  • Special attention has been paid to the optimization of package sizes
  • The CE marked products are compliant with EU regulatory requirements for in vitro diagnostic medical devices

Ordering Information: Not available in the U.S.A.

Specific Protein Applications for Indiko and Konelab Analyzers:

  • Albumin / Microalbuminuria
  • Apolipoprotein A1 & B
  • Complement C3 & C4
  • C-reactive protein
  • D-Dimer
  • Ferritin
  • HbA1c
  • IgA, G & M
  • Rheumatoid factors
  • Soluble transferrin receptor
  • Transferrin
Specifications
Certifications/ComplianceCE Marked
DescriptionHbA1c Pretreatment liquid; Kit size: 12 x 20mL
CE MarkerYes
Detectable AnalytesHbA1c
Sample TypeSerum
TypeReagent
Quantity12 x 20 mL
FormLiquid
Unit SizeEach