Indiko™ System Clinical Chemistry Reagents, Alkaline Phosphatase to Creatinine
Actual product may vary

Indiko™ System Clinical Chemistry Reagents, Alkaline Phosphatase to Creatinine

Standardize performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents, which offer best-in-class methodologies.
Have Questions?
Catalog NumberQuantityDetectable Analytes
9819504 x 25 mLAmylase
Catalog number 981950
Price (INR)
31,106.00
Each
Quantity:
4 x 25 mL
Detectable Analytes:
Amylase
Price (INR)
31,106.00
Each
Enhance your laboratory's performance with Thermo Scientific™ Indiko™ System Clinical Chemistry Reagents. These ready-to-use assays come fully validated and compliant, featuring automatic barcoding for efficient process and reordering checks. System reagents provide timely and accurate information, helping to ensure results you can stand behind.
  • Indiko™ applications utilize carefully selected methods according to IFCC and other widely used recommendations
  • Highly developed Indiko analyzers are integrated systems for routine clinical chemistry and specialty testing
  • Fully validated applications in accordance with the applicable EU regulatory requirements for IVD products permit holistic quality assurance in your lab
  • Clinical Chemistry methods are provided with system specified multi-calibrators and -controls with traceability to international reference materials
Ordering Information:

Not available in the U.S.A., except Glucose Hexokinase.

Clinical Chemistry Applications for Indiko Analyzers:
  • Albumin
  • ALP
  • ALT
  • Ammonia
  • Amylase
  • AST
  • Bile acids TBA
  • Bilirubin direct and total
  • Calcium
  • Copper
  • Cholesterol
  • HDL and LDL Cholesterol
  • Cholinesterase
  • Bicarbonate
  • Creatine Kinase
  • CK-MB
  • Creatinine
  • Gamma-GT
  • Glucose
  • G6P-DH
  • Iron
  • Lactate
  • LDH
  • Lipase
  • Magnesium
  • Phosphorus
  • Total Protein
  • Triglycerides
  • U/CSF Protein
  • Urea
  • Uric Acid
  • Zinc
Specifications
Certifications/ComplianceCE Marked
CE MarkerYes
ConcentrationReagent A: ≥ 2000 U/l alpha-Glucosidase, 62.5 mmol/l NaCl, 12.5 mmol/l MgCl2, < 0.1% NaN3, 0.1 mol/l Good's buffer, pH 7.15; Reagent B: 8.5 mmol/l EPS-G7, < 0.1% NaN3, 0.1 mol/l Good's buffer, pH 7.15
Detectable AnalytesAmylase
Sample TypeSerum, Plasma, Urine
FormulationReady for use
For Use With (Application)Indiko
Includes4 vials of 20 mL Reagent A; 4 vials of 5 mL Reagent B
Packaging TypeKit
Quantity4 x 25 mL
FormLiquid
Unit SizeEach
Contents & Storage
Stable at 2 to 8 °C until the expiration date stated on the label