Calcium (Ca) System Reagents, 350 tests
Actual product may vary
Calcium (Ca) System Reagents, 350 tests
Calcium (Ca) System Reagents, 350 tests
Calcium (Ca) System Reagents, 350 tests
Ready-to-use kits for rapid, cost-effective environmental analysis. Thermo Scientific™ Gallery™ system reagents and assay kits are ready to go.
Have Questions?
Catalog NumberQuantity
9843743 x 20 mL
Catalog number 984374
HSN 38229090
Price (INR)
4,743.00
Each
Quantity:
3 x 20 mL
Price (INR)
4,743.00
Each
Save time by eliminating reagent preparation with the Thermo Scientific™ Gallery™ system reagents for environmental testing, such as water (clean water, wastewater, soil and sludge digests, effluents, and saline) and industrial (process water) analysis. With over 40 ready-to-use Gallery system reagents for environmental analysis, the kits include 1 to 3 reagents in 20mL barcoded vials, allowing the necessary identification data, such as lot and expiration date, to be read automatically by the discrete analyzer's barcode reader. Automated dilutions make it possible to apply methods for a wide range of analyte concentrations and the test applications are based on recommended standard water and wastewater methods used in most countries. Save time and reduce errors by simplifying your analysis with the Gallery™ system reagents for environmental analysis.
Highlights:
  • All test applications are based on recommended standard methods in water and wastewater analysis of different countries
  • Kit structure includes 1 to 3 reagents in 20 mL vials
  • Barcoded vials are simple and fast to use
  • All necessary data, such as lot and expiration date, are automatically read
  • Automated dilutions make it possible to apply methods for a wide range of analyte concentrations
  • Types of matrices analyzed include clean water, wastewater, soil and sludge digests, effluents, saline and process water
Specifications
TypeCalcium System Reagent
Sample TypeWater Sample, Soil Sample
DescriptionCalcium (Ca) system reagents, 350 tests
DetectsCalcium (Ca)
No. of Tests350
Test Range0 to 1000
Quantity3 x 20 mL
Interference Filter660
Unit SizeEach