Product Image
Thermo Scientific™

Insulin MSIA Kit without LC Column

Simplify your affinity purification workflow with the Thermo Scientific™ InsuQuant™ Mass Spectrometric Kit, an exclusive pre-analytical solution designed to simultaneously detect, differentiate, and quantify endogenous and exogenous insulin types.
Have Questions?
Catalog number 991INSK96
Price (INR)
-
Simplify your affinity purification workflow with the Thermo Scientific™ InsuQuant™ Mass Spectrometric Kit, an exclusive pre-analytical solution designed to simultaneously detect, differentiate, and quantify endogenous and exogenous insulin types. Through the usage of proprietary microcolumn technology, the InsuQuant Mass Spectrometric Kit ships ready-to-use, containing the reagents and consumables needed to perform the affinity purification of insulin prior to liquid chromatography-mass spectrometry (LC-MS).
Simplify your bioanalysis workflow by having ready-to-use reagents and consumables for the qualitative and quantitative analysis of insulin at your fingertips.

The InsuQuant Mass Spectrometric Kit includes:
  • MSIA Insulin D.A.R.T.’S
  • Insulin Standard
  • Wash Buffer
  • Liquid Chromatography Column
  • Sample Dilution Buffer

The InsuQuant Mass Spectrometric Kit is compatible with semi-automated and high-throughput liquid handling equipment, which enables high sensitivity, low background, and reproducible results. The InsuQuant Mass Spectrometric Kit is best used with the following:
  • Thermo Scientific™ Versette™ Automated Liquid handler
  • Thermo Scientific™ Novus i™ electronic handheld pipette
Specifications
AntigenInsulin
Assay Range15 pM - 960 pM
Certifications/ComplianceFor Research Use Only
DescriptionInsulin MSIA Kit
Detection LimitLLOQ < 90 pg/mL; ULOQ > 5,576 pg/mL
Detection MethodMass spectrometry
For Use With (Application)For use with micro-scale analyte purification prior to mass spectrometric detection. All components are single use only. For research use only.
IncludesInsulin standard, internal reference standard, wash buffer, sample dilution buffer, elution solvent, Insulin MSIA DARTS
Length (Metric)185 mm
Product TypeConsumable Kit
Quantity96 Tests/Kit
Sample TypeBiological Samples
Sample Volume25 to 500 μL
Sensitivity25 pg/mL to 11,520 pg/mL
Shelf Life12 Months
Total Assay Time80 min.
Width (Metric)258 mm
Cross ReactivityInsulin analogues
Unit SizeCase of 1