Product Image
Thermo Scientific™

Insulin MSIA Kit with LC Column

Simplify your affinity purification workflow with the Thermo Scientific™ InsuQuant™ Mass Spectrometric Kit, an exclusive pre-analytical solution designed to simultaneously detect, differentiate, and quantify endogenous and exogenous insulin types.
Have Questions?
Catalog number 991INSK96C
Price (INR)
-
Simplify your affinity purification workflow with the Thermo Scientific™ InsuQuant™ Mass Spectrometric Kit, an exclusive pre-analytical solution designed to simultaneously detect, differentiate, and quantify endogenous and exogenous insulin types. Through the usage of proprietary microcolumn technology, the InsuQuant Mass Spectrometric Kit ships ready-to-use, containing the reagents and consumables needed to perform the affinity purification of insulin prior to liquid chromatography-mass spectrometry (LC-MS).
Simplify your bioanalysis workflow by having ready-to-use reagents and consumables for the qualitative and quantitative analysis of insulin at your fingertips.

The InsuQuant Mass Spectrometric Kit includes:
  • MSIA Insulin D.A.R.T.’S
  • Insulin Standard
  • Wash Buffer
  • Liquid Chromatography Column
  • Sample Dilution Buffer

The InsuQuant Mass Spectrometric Kit is compatible with semi-automated and high-throughput liquid handling equipment, which enables high sensitivity, low background, and reproducible results. The InsuQuant Mass Spectrometric Kit is best used with the following:
  • Thermo Scientific™ Versette™ Automated Liquid handler
  • Thermo Scientific™ Novus i™ electronic handheld pipette
Specifications
AntigenInsulin
Assay Range15 pM - 960 pM
Certifications/ComplianceFor Research Use Only
Cross ReactivityInsulin analogues
DescriptionInsulin MSIA Kit
Detection LimitLLOQ < 90 pg/mL; ULOQ > 5,576 pg/mL
Detection MethodMass spectrometry
IncludesInsulin standard, internal reference standard, wash buffer, LC column, sample dilution buffer, elution solvent, Insulin MSIA DARTS
Length (Metric)185 mm
Quantity96 Tests/Kit
Sample TypeBiological Samples
Sample Volume25 to 500 μL
Sensitivity25 pg/mL to 11,520 pg/mL
Total Assay Time80 min.
Width (Metric)258 mm
For Use With (Application)For use with micro-scale analyte purification prior to mass spectrometric detection. All components are single use only. For research use only.
Shelf Life12 Months
Product TypeConsumable Kit
Unit SizeCase of 1