AlgiMatrix™ 6-Well, 3D Culture System, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Change view
Catalog Number
Quantity
No. of Wells
A1098201
1 x 6 well plate
6
A1098202
4 x 6 well plates
6
2 Options
Catalog number A1098201
Price (INR)
-
Quantity:
1 x 6 well plate
No. of Wells:
6
AlgiMatrix™ 3D Culture System 6 well plate is a user-friendly, animal –free bioscaffold available for the development of high-fidelity cell culture models that are more predictive of disease states and drug responses. AlgiMatrix™ sponges are formulated using pharmaceutical-grade raw material from brown seaweed. Its pure, chemically defined, and nontoxic macro porous structure improved lot-to-lot consistency, superior cell loading and excellent nutrient delivery, without damage to cells. The AlgiMatrix™ sponge is biodegradable and is stable at room temperature.
The AlgiMatrix™ 3D Culture System is suitable for a broad range of procedures; including Multicellular tumor spheroid assays (MCTS), hepatocyte and cardiomyocyte organogenesis studies, co-culture studies, and high-throughput drug screening assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes1 x 6 well AlgiMatrix Plate and 50 mL Firming Buffer
LidWith Lid
Product TypeMicroplate
Purity or Quality GradeCell Culture Grade
Quantity1 x 6 well plate
Shipping ConditionRoom Temperature
ColorClear
No. of Wells6
Product LineAlgimatrix
SterilitySterile
Surface TreatmentAlgiMatrix 3D Culture System
Well ShapeFlat Bottom (F-well)
Unit SizeEach
Contents & Storage
Contains 1 x 6 well AlgiMatrix™ plate and 50 ml firming buffer. Store at room temperature, 15-30° C
Frequently asked questions (FAQs)
How stable is AlgiMatrix Matrix at room temperature without degradation?
AlgiMatrix Matrix is stable for 12 months unopened. Once opened, the entire plate needs to be used.
Why can Gibco Geltrex Matrix be used for adhesion-related research, as opposed to Gibco AlgiMatrix 3D Culture System which cannot?
Geltrex Matrix is a basement membrane extract that contains collagen IV, laminin, and other matrix proteins and growth factors that are required for cell adhesion. AlgiMatrix Matrix contains only alginate which does not facilitate cell adhesion. There is nothing in the alginate sponges for the cells to interact with.
Can other cell viability reagents be used with AlgiMatrix Matrix?
Other than the Invitrogen alamarBlue Cell Viability Reagent, AlgiMatrix Matrix is also compatible with Invitrogen PrestoBlue Cell Viability Reagent and Invitrogen CyQuant Direct Cell Proliferation Assay. The protocol is almost the same as the 2D cell culture, but it needs the blank AlgiMatrix Matrix as a blank control.
How important is calcium in the AlgiMatrix firming and dissolving buffers?
The firming buffer works by adding calcium to the sponge, making it stiffer. The dissolving buffer works by removing calcium from the sponge, making it softer.
Is there any assay variability when using AlgiMatrix Matrix?
3D cell culture with AlgiMatrix Matrix approaches a more physiological state in terms of cell growth and behavior, but along with that comes the higher variability typically associated with more complex systems. You may need to do more replicates or more controls.