CHO-S Cells (cGMP banked) and Media Kit
Gibco™
CHO-S Cells (cGMP banked) and Media Kit
Gibco™ CHO-S Cells (cGMP Banked) and Media Kit have been developed for the growth of Chinese Hamster Ovary (CHO) cells and expression of recombinant proteins in suspension culture.
Have Questions?
Catalog NumberQuantity
A11557011 Kit
Catalog number A1155701
Price (INR)
1,526,242.00
Each
Quantity:
1 Kit
Price (INR)
1,526,242.00
Each
Gibco® CHO-S Cells (cGMP Banked) and Media Kit have been developed for the growth of Chinese Hamster Ovary (CHO) cells and expression of recombinant proteins in suspension culture. Parental CHO-S cells have been produced, banked and tested to meet cGMP quality standards. The cells have been adapted to CD CHO Medium for serum free suspension growth. The CD CHO Medium is made from animal origin free and chemically defined components. It contains no proteins, hydrolysates, or components of unknown composition. The medium is formulated without L-glutamine for greater stability, and without phenol red to minimize potential for estrogen-like effects.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ClassificationLow-Protein, Serum-Free
Media RecommendationCD CHO Medium
No. of Cells1 x 107
Product LineGibco
Quantity1 Kit
Cell LineCHO-S Cells
Culture TypeSuspension Cell Culture
SpeciesHamster
Unit SizeEach
Contents & Storage
CHO-S Cells (cGMP banked) and Media Kit (SKU A1155701) contains:
CD CHO Medium 1000 mL- 10743-029, store in the dark at 2-8°C
>CHO-S Cells (cGMP banked) 1 vial (> 1x10^7 cells) - A11364-01 (see limited label license), store in liquid nitrogen
>L-glutamine-200mM (100X) 100 mL - 25030-081 or 25030-024 (CE marked version for Europe only), store at -20°C
Frequently asked questions (FAQs)
What level of product quality is seen with the Freedom CHO-S Kit?
After execution and payment for a Commercial Production License, a Cell Line Documentation Package (>200 pages, and specific to the lot of cells purchased), is provided, which includes the entire lineage history of the cells, starting at receipt of the initial vial at Thermo Fisher Scientific through delivery to the purchaser, as well as, all testing reports.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
How can I learn more about commercial licensing of Freedom kits, ExpiCHO-S Cells (cGMP banked), CHO-S Cells (cGMP banked), and CHO DG44 Cells (cGMP banked)?
To inquire about a Commercial Production or Service License, please email: outlicensing@thermofisher.com.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
What is the term of the license agreement for ExpiCHO-S Cells (cGMP banked), CHO-S Cells (cGMP banked), and CHO DG44 Cells (cGMP banked)?
Licensing rights are granted in perpetuity for the Commercial Production License. The term of the Commercial Service License is negotiated between the parties.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Is there a pull-through royalty on final products or an annual maintenance fee for using ExpiCHO-S Cells (cGMP banked), CHO-S Cells (cGMP banked), and CHO DG44 Cells (cGMP banked)??
Only a one-time license fee is required for a Commercial Production License, and it covers use during the clinical trial process and commercialization of an unlimited number of molecules.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.
Are separate licenses required for ExpiCHO-S Cells (cGMP banked), CHO-S Cells (cGMP banked), and CHO DG44 Cells (cGMP banked)?
Yes. Each cell line requires a separate Commercial Production or Service License.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.