BODIPY™ FL ATP (Adenosine 5'-Triphosphate, BODIPY™ FL 2'-(or-3')-O-(N-(2-Aminoethyl)Urethane), Trisodium Salt), 5 mM in buffer
BODIPY FL adenosine 5'-triphosphate and BODIPY FL guanosine 5'-triphosphate (G-12411) are composed of a BODIPY FL fluorophore attached to theRead more
Have Questions?
Catalog Number
Quantity
A12410
also known as A-12410
100 μL
Catalog number A12410
also known as A-12410
Price (INR)
-
Quantity:
100 μL
BODIPY FL adenosine 5'-triphosphate and BODIPY FL guanosine 5'-triphosphate (G-12411) are composed of a BODIPY FL fluorophore attached to the 2' or 3' position of the ribose ring via a linker. Interactions between the fluorophore and the purine base are evident from the spectroscopic properties of these nucleotide analogs. The fluorescence quantum yield of BODIPY FL GTP is only about one-tenth that of BODIPY FL ATP; however, this yield may be enhanced upon binding of BODIPY FL GTP to GTP-binding proteins.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeBODIPY™ FL
Product TypeFL ATP
Quantity100 μL
Shipping ConditionWet Ice
Concentration5 mM
Product LineBODIPY
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.
Citations & References (14)
Citations & References
Abstract
Hexokinase inhibits flux of fluorescently labeled ATP through mitochondrial outer membrane porin.
Authors:Perevoshchikova IV, Zorov SD, Kotova EA, Zorov DB, Antonenko YN,
Journal:FEBS Lett
PubMed ID:20412805
'Mitochondrial function requires maintaining metabolite fluxes across the mitochondrial outer membrane, which is mediated primarily by the voltage dependent anion channel (VDAC). We applied fluorescence correlation spectroscopy (FCS) to study regulation of the VDAC functional state by monitoring distribution of fluorescently labeled ATP (BODIPY-FL-ATP) in isolated intact rat liver and ... More
Evidence for major structural changes in subunit C of the vacuolar ATPase due to nucleotide binding.
Authors:Armbrüster A, Hohn C, Hermesdorf A, Schumacher K, Börsch M, Grüber G
Journal:FEBS Lett
PubMed ID:15792803
The ability of subunit C of eukaryotic V-ATPases to bind ADP and ATP is demonstrated by photoaffinity labeling and fluorescence correlation spectroscopy (FCS). Quantitation of the photoaffinity and the FCS data indicate that the ATP-analogues bind more weakly to subunit C than the ADP-analogues. Site-directed mutagenesis and N-terminal sequencing of ... More
ATP modulation of excitatory synapses onto interneurons.
Authors:Khakh BS, Gittermann D, Cockayne DA, Jones A
Journal:J Neurosci
PubMed ID:12917379
Inhibitory interneurons play important roles in neuronal circuits, but the synaptic mechanisms that regulate excitatory input onto interneurons remain to be fully understood. We show that ATP-gated presynaptic P2X2 channels facilitate excitatory transmission onto stratum radiatum interneurons but not onto CA1 pyramidal neurons. ATP released endogenously during carbachol-induced oscillations facilitates ... More
Detection of adenylyl cyclase activity using a fluorescent ATP substrate and capillary electrophoresis.
Authors:Cunliffe JM, Sunahara RK, Kennedy RT
Journal:Anal Chem
PubMed ID:16536404
A capillary electrophoresis laser-induced fluorescence (CE-LIF) assay was developed for detection of adenylyl cyclase (AC) activity using BODIPY FL ATP (BATP) as substrate. In the assay, BATP was incubated with AC and the resulting mixture of BATP and enzyme product (BODIPY cyclic AMP, BcAMP) separated in 5 min by CE-LIF. ... More
Tannin Supplementation Improves Oocyte Cytoplasmic Maturation and Subsequent Embryo Development in Pigs.