Annexin V Conjugates for Apoptosis Detection
Invitrogen™

Annexin V Conjugates for Apoptosis Detection

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/EmissionFlow Cytometer Laser LinesConjugate
A13202578/603532, 561Alexa Fluor 568
A13201495/519488Alexa Fluor 488
A13199494/518488FITC
A13203590/617532Alexa Fluor 594
A13204Biotin-X
A23202346/442UVAlexa Fluor 350
A23204650/665633-637Alexa Fluor 647
A35108555/565532, 561Alexa Fluor 555
A35109679/702633-637Alexa Fluor 680
A35110650/660633-637APC (Allophycocyanin)
A35111565/578488, 532, 561PE
A35122410/455405Pacific Blue
Catalog number A13202
Price (INR)
26,750.00
Online offer
Ends: 31-Dec-2026
44,583.00
Save 17,833.00 (40%)
Each
Excitation/Emission:
578/603
Flow Cytometer Laser Lines:
532, 561
Conjugate:
Alexa Fluor 568
Price (INR)
26,750.00
Online offer
Ends: 31-Dec-2026
44,583.00
Save 17,833.00 (40%)
Each
Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.
Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorOrange-Red
DescriptionAnnexin V, Alexa Fluor 568 conjugate
Excitation/Emission578/603
Flow Cytometer Laser Lines532, 561
For Use With (Equipment)Flow Cytometer
No. of Reactions100
Product TypeAnnexin V conjugate
Quantity500 μL
Shipping ConditionWet Ice
ConjugateAlexa Fluor 568
Unit SizeEach
Contents & Storage
Contains 1 vial of annexin V, Alexa Fluor 568 conjugate. Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

I want to study apoptosis using an Annexin V conjugate, but with adherent cells via microscopy instead of flow cytometry. Can this be done?

It has been done, but we don‘t recommend it. Both healthy cells and apoptotic cells possess phosphatidylserine on the cell surface, which can be detected with Annexin V, but apoptotic cells have significantly more of it. You can easily tell the difference between these two populations with flow cytometry, because flow cytometers are more sensitive and have a higher throughput. But with a microscope, you cannot always tell the difference, especially for adherent cells. Instead, for microscopy, we recommend a different technique, such as detecting caspases with CellEvent Caspase Detection Reagents.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When should I use Annexin V instead of an Invitrogen CellEvent Caspase Detection Reagent?

Choose Annexin V to detect phosphatidylserine exposure at the plasma membrane, and choose Invitrogen CellEvent Caspase Detection Reagents to detect activated caspases and nuclear fluorescence. The methods measure different apoptotic events and should not be treated as interchangeable.

I trypsinized my adherent cells and labeled with annexin V, and now my flow data is showing a high percentage of apoptotic cells even for control, untreated cells. What is the problem?

Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. Allow the cells to recover for about 30 minutes in optimal cell culture conditions and medium after trypsinizing/scraping so that they can recover their membrane integrity before staining. For lightly adherent cell lines, such as HeLa and NIH 3T3, another option is to use non-enzyme treatments like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I detect annexin V staining in an imaging assay?

Annexin V staining is not typically used in imaging experiments; it is a better reagent for flow cytometry analysis. All cells will stain to some extent, so it can be difficult to distinguish a relatively bright annexin V-stained cell from a dimmer non-apoptotic cell. Caspase activation, detected using our CellEvent Caspase 3/7 or Image-iT LIVE Caspase detection kits, is a better method for detecting apoptosis in an imaging assay.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When should I stain adherent cells with annexin V for flow cytometric analysis? Before or after I trypsinize them?

Trypsinize first and then allow the cells to recover about 30 minutes in optimal cell culture conditions and medium before staining with annexin V conjugates. Trypsinization or mechanical scraping of cells temporarily disrupts the plasma membrane, allowing for annexin V to bind phosphatidylserine on the cytoplasmic surface of the cell membrane and thus leading to false positive staining. For lightly adherent cell lines such as HeLa and NIH 3T3, you could use a less harsh (non-enzymatic) dissociation product like Gibco Cell Dissociation Buffer (Cat. No. 13151014).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (14)

Citations & References
Abstract
Quantitation of microparticles released from coated-platelets.
Authors:Dale GL, Remenyi G, Friese P,
Journal:J Thromb Haemost
PubMed ID:16102115
'Dual agonist stimulation of platelets with thrombin and convulxin results in generation of coated-platelets, a sub-population of cells known formerly as COAT-platelets (collagen and thrombin). Coated-platelets retain several procoagulant proteins on their surface and express phosphatidylserine (PS). In this report, we utilize a new methodology to demonstrate that coated-platelets also ... More
Bacterium-generated nitric oxide hijacks host tumor necrosis factor alpha signaling and modulates the host cell cycle in vitro.
Authors:Mocca B, Wang W,
Journal:J Bacteriol
PubMed ID:22636782
'In mammalian cells, nitric oxide (NO·) is an important signal molecule with concentration-dependent and often controversial functions of promoting cell survival and inducing cell death. An inducible nitric oxide synthase (iNOS) in various mammalian cells produces higher levels of NO· from l-arginine upon infections to eliminate pathogens. In this study, ... More
Accumulation of glycosphingolipids in Niemann-Pick C disease disrupts endosomal transport.
Authors:te Vruchte D, Lloyd-Evans E, Veldman RJ, Neville DC, Dwek RA, Platt FM, van Blitterswijk WJ, Sillence DJ
Journal:J Biol Chem
PubMed ID:15078881
'Glycosphingolipids are endocytosed and targeted to the Golgi apparatus but are mistargeted to lysosomes in sphingolipid storage disorders. Substrate reduction therapy utilizes imino sugars to inhibit glucosylceramide synthase and potentially abrogate the effects of storage. Niemann-Pick type C (NPC) disease is a disorder of intracellular transport where glycosphingolipids (GSLs) and ... More
Phosphatidylserine exposure in B lymphocytes: a role for lipid packing.
Authors:Elliott JI, Sardini A, Cooper JC, Alexander DR, Davanture S, Chimini G, Higgins CF,
Journal:Blood
PubMed ID:16684961
'Plasma membrane lipids are usually distributed asymmetrically, with phosphatidylserine (PS) confined to the inner leaflet. PS exposure at the outer leaflet occurs early in apoptosis, but it is also constitutive on some nonapoptotic cell populations where it plays a role in cell signaling. How PS is transported ("flopped") to the ... More
A ribonucleotide reductase homolog of cytomegalovirus and endothelial cell tropism.
Authors:Brune W, Ménard C, Heesemann J, Koszinowski UH
Journal:Science
PubMed ID:11209080
Human cytomegalovirus infects vascular tissues and has been associated with atherogenesis and coronary restenosis. Although established laboratory strains of human cytomegalovirus have lost the ability to grow on vascular endothelial cells, laboratory strains of murine cytomegalovirus retain this ability. With the use of a forward-genetic procedure involving random transposon mutagenesis ... More