Expi293™ Expression System Kit
Gibco™

Expi293™ Expression System Kit

Designed with transient expression technology for rapid and ultra high-yield protein production from mammalian cells.
Have Questions?
Catalog NumberQuantity
A146351 Kit
Catalog number A14635
Price (INR)
175,810.00
Online offer
Ends: 31-Dec-2026
251,157.00
Save 75,347.00 (30%)
Each
Quantity:
1 Kit
Price (INR)
175,810.00
Online offer
Ends: 31-Dec-2026
251,157.00
Save 75,347.00 (30%)
Each

The Expi293 Expression System is a complete, scalable platform for rapid, high-yield transient protein production from mammalian HEK293 cells. The system includes: high-density Expi293F cells in Expi293 Expression Medium and ExpiFectamine 293 transfection reagent optimized with enhancers to maximize protein production and reproducibility.

  • Complete kit—contains all components needed for efficient, recombinant protein expression with easy-to-follow culture and transfection protocols
  • Up to 1 g/L of protein—2- to 10-fold higher protein yields than previous generation transient expression systems, such as the FreeStyle 293 Expression System
  • Rapid production—rapid expression of recombinant proteins in just 2–7 days for a wide variety of protein formats from integral membrane proteins to monoclonal antibodies
  • High density cultures—high-density Expi293F cultures yield significantly more protein-expressing cells per mL, boosting expression efficiency
  • Native folding and mammalian post-translational modifications—proteins are expressed from human HEK293 cells resulting in a high degree of glycosylation and quality
  • Scalable expression from <1 mL to >10 L—easily scalable and automation friendly from 96 deepwell plates for high throughput applications to multi-liter bioreactor platforms

The components included in the Expi293 Expression System Kit are: two vials of frozen Expi293F cells, one liter of Expi293 Expression Medium, one ExpiFectamine 293 Transfection kit sufficient to transfect up to 1 liter of final culture, Opti-MEM I Reduced Serum Medium complexation buffer, and an antibody-expressing positive control vector. Expi293 Transfection kits are also available in large-scale formats for transfecting up to 10 L and 50 L of Expi293 cells. Custom formats and volumes are also available upon request.

Engineered Expi293F cell lines are available for a range of structural biology applications, including Cryo-EM, NMR, and X-ray crystallography. Available individually or in kits.

  • Expi293F GnTI- Cells (Cat. No. A39240)
  • Expi293F Inducible Cells (Cat. No. A39241)
  • Expi293F Inducible GnTI- Cells (Cat. No. A39242)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineExpi293F™
Delivery TypeTransfection
Expression StabilityTransient
Expression SystemMammalian
For Use With (Application)Protein Expression
Key FunctionsProtein Production, Constitutive Expression
No. of Reactions33
Product LineExpi293
Product TypeExpression System Kit
Protein TagUntagged
Quantity1 Kit
System TypeExpi293™ Expression System
Cell Type293-derived Cells
PromoterCMV
Transfection TechniqueLipid-based Transfection
Unit SizeEach
Contents & Storage
2 vials frozen Expi293F cells
1 bottle 1000 mL Expi293 Expression Medium
1 ExpiFectamine 293 transfection kit for 1 L of culture
1 vial Antibody Expressing Positive Control Vector
1 bottle 100 mL Opti-MEM Reduced Serum Medium

Store all components at +2°C to +8°C, except frozen cells are stored in liquid nitrogen

Frequently asked questions (FAQs)

I do not have Expi293F cells. Can I use a different 293 cell line with the Expi293 Expression System?

Most 293 cell lines can be adapted directly from conventional serum-containing or other serum-free media into Expi293 Expression Medium by either using the Direct Adaptation method or Sequential Adaptation method, described in the manual.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Can I use the Expi293 Expression System for scalable production of membrane proteins?

Please refer to the Application Note (http://www.thermofisher.com/content/dam/LifeTech/global/life-sciences/CellCultureandTransfection/pdfs/Expi293Adaption-MembraneProTech_AppNoteGlobal_FHR.pdf) for a protocol for adapting the MembranePro kit for use with the Expi293 System.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Do you have a protocol for adapting the Expi293 Expression System for use in the WAVE Bioreactor System?

Please see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.0358.file.dat/PG1391-PJ6257-CO28716-Expi293-10L-Wave-Bag-App-Note-Global_FHR.pdf) for a method for the adaptation of the Expi293 Expression System for use in the WAVE Bioreactor System.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Can I express proteins in a 96-well or other multiwell format using the Expi293 Expression System?

Yes, see the Application Note (http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.28891.file.dat/expi293-microtiter-plates-co25793.pdf) on using 96-well microtiter plates with the Expi293 Expression System.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

Do you have any tips for optimizing protein yield using the Expi293 Expression System?

Please refer to the Application Note for tips on optimizing protein yield using the Expi293 Expression System (http://www.thermofisher.com/content/dam/LifeTech/migration/files/proteins-expression-isolation-analysis/pdfs.par.61153.file.dat/expi293-protein-yield-co25763.pdf).

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.