TaqMan™ hPSC Scorecard™ Panel, 384-well
Applied Biosystems™

TaqMan™ hPSC Scorecard™ Panel, 384-well

The Applied Biosystems™ TaqMan™ hPSC Scorecard™ Panel 384w enables verification of pluripotency and determination of lineage bias for both humanRead more
Have Questions?
Catalog NumberQuantity
A158701 x 384-well plate
Catalog number A15870
Price (INR)
-
Quantity:
1 x 384-well plate
The Applied Biosystems™ TaqMan™ hPSC Scorecard™ Panel 384w enables verification of pluripotency and determination of lineage bias for both human ES and iPS cell lines. The 384-well plate contains four sets of 94 predefined TaqMan™ Gene Expression assays (including endogenous controls) dried-down in the wells. Master mix is not included with this panel. For a kit that combines the panel with master mix, see TaqMan™ hPSC Scorecard™ Kit 384w. This product is also available in a 2 x 96w Fast format.

• Assess the gene signature of four samples in one simple experiment
• Compare results to a reference standard with the accompanying hPSC Scorecard™ Analysis Software
• Identify lines with trilineage differentiation potential
• Evaluate inductive differentiation specified to one germ layer
• Access validated content for increased confidence in your results

Assess an Entire Gene Signature in One Experiment
The TaqMan™ hPSC Scorecard™ Panel helps save time by offering pre-plated assays in a stable and convenient dried-down format. Simply add TaqMan™ Master Mix to your cDNA of interest and transfer to the 384-well plate with a multichannel pipette.

Compare Results to a Reference Standard
The TaqMan™ hPSC Scorecard™ Panel includes access to proprietary cloud-based analysis software that allows you to view and export graphs and tables of results, as well as generate reports. The software is compatible with seven Applied Biosystems™ qRT-PCR systems and is available at no additional charge.

Identify Lines with Trilineage Differentiation Potential
Include randomly differentiated embryoid body (EB) samples (differentiated for at least 7 days using standard EB methods) in your analysis and determine whether your lines are biased toward one or more of the three germ layers (endoderm, mesoderm, ectoderm).

Evaluate Inductive Differentiation Specified to One Germ Layer
The panel includes over twenty markers for each of the three germ layers that have been shown to respond as expected to directed differentiation in monolayer culture. Quickly evaluate time course data, culture conditions and media formulations for your germ layer of interest with this one panel.

Access Validated Content
The panel contents are based on published work (Bock et al., Cell 144, 439–452, 2011) and were validated against multiple human ES and iPS lines.

What You Get
The TaqMan™ hPSC Scorecard™ Panel 384w includes:

• One 384-well plate of 94 TaqMan™ Gene Expression assays (4 gene sets per plate)
• One optical plate cover
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™ 12k Flex, ViiA™ 7 System
Format384-well plate
No. of Reactions384 Reactions
Product LineTaqMan
Quantity1 x 384-well plate
Research CategoryStem Cell Research
SpeciesHuman
Detection MethodPrimer-probe
For Use With (Application)Stem Cell Analysis
FormDried Down
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contents: One 384-well plate with four replicates of the 96-assay panel and one optical cover

Store at room temperature.

Frequently asked questions (FAQs)

Will I need to update my TaqMan hPSC Scorecard Analysis Software?

No software update is required. Because the hPSC Scorecard Analysis Software is cloud based, you will always have access to the most up-to-date version of the software every time you log in.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center

How do I export Ct values in the TaqMan hPSC Scorecard Analysis Software?

The only way to view the Ct values in the current version is through the expression plot. We expect the software's Excel export to include the gene names and Ct values in a future version of the software.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center

I'm using the TaqMan hPSC Scorecard Analysis Software. How are the scores calculated?

A proprietary algorithm compares the Ct values for each marker set to the values in the reference database and calculates the score based on how well the expression correlates. In general, scores close to 0 indicate comparable expression to that of the reference standard using undifferentiated cells. Scores higher than 1 indicates up regulation relative to undifferentiated cells and less than -1 indicate down regulation relative to undifferentiated cells.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center

I'm using the TaqMan hPSC Scorecard Analysis Software. What lines were used to generate the reference standard?

Thirteen pluripotent stem cell lines were used to generate the reference standard including:

  • H9 ESC P28
  • iPS BS3C P35
  • iPS18C P29
  • HUES9 P28
  • HUES13 P56
  • HUES28 P28
  • HUES44 P25
  • HUES48 P21
  • HUES49 P21
  • HUES53 P24
  • HUES63 P46
  • HUES64 P30
  • HUES65 P29


Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center

I'm using the TaqMan hPSC Scorecard Analysis Software. My sample has a red flag on it, or is not showing any results. What does that mean?

The hPSC Scorecard Analysis Software performs a set of quality control checks upon data import to ensure the quality of the results.
You may see one of the following red flag warnings:

  • Internal positive control: The expression levels of the housekeeping genes were low, signifying that there was an issue with the qRT-PCR. The sample is excluded from analysis and must be repeated.
  • Bad Rox: The ROX dye is used as a passive reference during the qPCR. If the ROX dye is not detected, it is suggestive that some of the assay wells may not have received the proper amount of Master Mix. This may explain why a particular assay is not showing the anticipated gene expression level.
  • Sendai Virus Detected (SEV): The hPSC Scorecard Panel includes a control to look for Sendai virus expression. A flag will appear if a Ct value of greater than 30 is observed for this control. On some rare occasions, the SEV oligonucleotides will cross react with other genes resulting in a false SEV flag in differentiated cells. If you see this flag, and did not anticipate Sendai virus detection, you may want to repeat the sample or look at the amplification plot (In the 96 well plate: well B9; in the 384 well plate: wells D3, D9, D15, and D21) to see if the automated Ct value call is real, or just an error.
  • Insufficient Data: The analysis software requires a minimum number of genes to be amplified in each the four categories (i.e., self-renewal (“pluri”), ectoderm, mesoderm, and endoderm) to accurately analyze your data. If the minimum number of genes in each category does not amplify, or if data has been omitted by the user, the Insufficient Data flag will appear and the sample is excluded from analysis. The sample should be repeated.


Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center

Citations & References (20)

Citations & References
Abstract
Characterizing Pluripotent Stem Cells Using the TaqMan(®) hPSC Scorecard (TM) Panel.
Authors:Fergus J, Quintanilla R, Lakshmipathy U,
Journal:
PubMed ID:25138722
'Rapid technological developments for the efficient generation of footprint-free induced pluripotent stem cells (iPSC) enabled the creation of patient-specific iPSC for downstream applications in drug discovery and regenerative medicine. However, the large number of iPSCs, generated from diverse genetic backgrounds using various methods and culture conditions, created a steep challenge ... More
Generation of iPSCs as a Pooled Culture Using Magnetic Activated Cell Sorting of Newly Reprogrammed Cells.
Authors:Yang W, Liu Y, Slovik KJ, Wu JC, Duncan SA, Rader DJ, Morrisey EE,
Journal:
PubMed ID:26281015
'Although significant advancement has been made in the induced pluripotent stem cell (iPSC) field, current methods for iPSC derivation are labor intensive and costly. These methods involve manual selection, expansion, and characterization of multiple clones for each reprogrammed cell sample and therefore significantly hampers the feasibility of studies where a ... More
Efficient Generation of Induced Pluripotent Stem and Neural Progenitor Cells From Acutely Harvested Dura Mater Obtained During Ventriculoperitoneal Shunt Surgery.
Authors:Cary WA, Hori CN, Pham MT, Nacey CA, McGee JL, Hamou M, Berman RF, Bauer G, Nolta JA, Waldau B,
Journal:
PubMed ID:26074438
'The dura mater can be easily biopsied during most cranial neurosurgical operations. We describe a protocol that allows for robust generation of induced pluripotent stem cells (iPSCs) and neural progenitors from acutely harvested dura mater. To generate iPSCs and neural progenitor cells from dura mater obtained during ventriculoperitoneal shunt surgery. ... More
Integrative Analyses of Human Reprogramming Reveal Dynamic Nature of Induced Pluripotency.
Authors:Cacchiarelli D, Trapnell C, Ziller MJ, Soumillon M, Cesana M, Karnik R, Donaghey J, Smith ZD, Ratanasirintrawoot S, Zhang X, Ho Sui SJ, Wu Z, Akopian V, Gifford CA, Doench J, Rinn JL, Daley GQ, Meissner A, Lander ES, Mikkelsen TS,
Journal:
PubMed ID:26186193
'Induced pluripotency is a promising avenue for disease modeling and therapy, but the molecular principles underlying this process, particularly in human cells, remain poorly understood due to donor-to-donor variability and intercellular heterogeneity. Here, we constructed and characterized a clonal, inducible human reprogramming system that provides a reliable source of cells ... More
Transcription factor binding dynamics during human ES cell differentiation.
Authors:Tsankov AM, Gu H, Akopian V, Ziller MJ, Donaghey J, Amit I, Gnirke A, Meissner A,
Journal:
PubMed ID:25693565
'Pluripotent stem cells provide a powerful system to dissect the underlying molecular dynamics that regulate cell fate changes during mammalian development. Here we report the integrative analysis of genome-wide binding data for 38 transcription factors with extensive epigenome and transcriptional data across the differentiation of human embryonic stem cells to ... More