LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit
LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit
The LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential androgenRead more
Have Questions?
Catalog NumberQuantity
A15878800 x 20 μL assays
Catalog number A15878
Price (INR)
220,248.00
Each
Quantity:
800 x 20 μL assays
Price (INR)
220,248.00
Each
The LanthaScreen™ TR-FRET Androgen Receptor Coactivator Assay Kit provides a sensitive and robust method for high-throughput screening of potential androgen receptor (AR) ligands as agonists of coactivator recruitment or antagonists of agonist-dependent coactivator recruitment. In both the agonist and antagonist modes, the homogeneous mix-and-read assay uses a rat AR ligand-binding domain (AR-LBD) tagged with hexahistidine and glutathione-S-transferase (GST) (also available separately), a terbium (Tb)-labeled anti-GST antibody, and a fluorescein-labeled coactivator peptide.
Agonist Mode
When the LanthaScreen TR-FRET Androgen Receptor Coactivator Assay is run in agonist mode (to identify agonist compounds), AR-LBD is added to ligand test compounds, followed by the addition of a mixture of fluorescein-coactivator peptide and Tb-anti-GST antibody. After room temperature incubation, the TR-FRET 520:495 nm emission ratio is calculated and used to determine the EC50 from a dose response curve of the compound. This ligand EC50 is a composite value representing the amount of ligand required to bind to receptor, effect a conformational change, and recruit coactivator peptide (see figure).
Antagonist Mode
When the LanthaScreen TR-FRET Androgen Receptor Coactivator Assay is run in antagonist mode (to identify antagonist compounds), AR-LBD is added to ligand test compounds, followed by addition of a mixture of DHT agonist, fluorescein-coactivator peptide, and Tb-anti-GST antibody. The concentration of DHT agonist used in this mode is the EC80 concentration as determined by first running the assay in agonist mode. An example of data produced in the antagonist mode is shown in the figure below.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Co-Factor Interaction Assay, Nuclear Receptor Assay
LigandAR
No. of Assays800 x 20 μL assays
Product TypeTR-FRET Androgen Receptor Coactivator Assay Kit
Quantity800 x 20 μL assays
Product LineLanthaScreen
Unit SizeEach
Contents & Storage
1 tube AR Ligand Binding Domain Recombinant Protein (store at -68 to -85°C)
1 tube Peptide (store at -5 to -30°C)
2 bottles TR-FRET Buffer (store at -5 to -30°C)
1 tube Tb-Anti-GST Ab (store at -5 to -30°C)
1 tube of DTT (store at -5 to -30°C)
Frequently asked questions (FAQs)
I'm using the TaqMan hPSC Scorecard Panel. How do I load the samples onto a 384-well plate if I only have a 16-channel pipette?
The tips of most 16-channel pipettes will align with every well in each column of the plate. However, if your cDNA reactions were set up in 8 wells of a 96-well plate or in 8-well PCR strips, additional sample will be required to compensate for the dead volume. When you insert 2 tips of the 16-channel pipette into 1 well, the tips can't reach the bottom of the well, resulting in a need for additional dead volume.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center
How does the LanthaScreen technology compare to other TR-FRET assay formats?
We performed a comparison between the LanthaScreen assay and other commercially available TR-FRET assays from 2 different suppliers for the PKC kinase target. Our data revealed that the assays performed comparably, but that the LanthaScreen assay was simpler to optimize and contained fewer components that required optimization. The LanthaScreen assay is a two component system, whereas the other assay formats utilize a trimolecular mechanism which is more time consuming to optimize and has added costs.
For my kinase assay, can I pre-mix the Tb-Ab and EDTA so that I can stop the kinase assay and begin detection with a single reagent addition?
Yes, this is possible depending on the concentrations of reagents used and the time for which they are mixed. We recommend developing and optimizing the assay by using separate reagent additions, because this method will work under the widest range of conditions. Once the assay is optimized, the performance of the assay using pre-mixed antibody and EDTA can be evaluated. We have successfully developed robust assays in which the antibody and EDTA were pre-mixed and then stored overnight at 4 degrees C prior to use the following day. A loss of signal intensity was observed in this case, however, by using the ratiometric readout, this effect was minimal.
Are the LanthaScreen reagents stable to interference from Mg2+, Mn2+, and EDTA?
The chelate is completely stable to Mg2+. The amount of Mn2+ or EDTA that the chelate can tolerate depends largely on how long they are mixed together and the combination of additives used in the reaction. If a reaction requires either Mg2+ or Mn2+ for activation, it is best to stop the reaction by adding an equimolar amount (or slight excess) of EDTA to chelate the metal ions present. This will then essentially eliminate any interference on the terbium chelate by EDTA or Mn2+. Regardless, when LanthaScreen assays are performed using a ratiometric readout (division of the acceptor signal by the donor signal), any interference caused by Mn2+ or EDTA is largely cancelled out.
What is the optimal and/or maximum distance for a Tb-fluorescein pair?
The Förster radius, the distance at which energy transfer efficiency is half-maximal, is around 50-angstroms for the terbiumÆ fluorescein pair. However, the Förster radius does not give a complete indication of energy transfer efficiency when using long lifetime fluorophores such as terbium chelates. When using terbium chelates, energy transfer efficiency is determined by the distance of closest approach between the donor and acceptor during the excited state lifetime of the donor. In many assay systems, such as those designed using antibodies or peptides, there is a large degree of conformational freedom that allows the donor and acceptor to approach one another, effectively enhancing the FRET signal. Additionally, it is important to note that as the donor/acceptor pair approach one another and the efficiency of energy transfer increases, the fluorescent lifetime decreases to a comparable extent. From a practical standpoint, this means that when energy transfer is extremely efficient, FRET cannot be measured in time-resolved mode (because the energy transfer is complete before the measurement is made). This is another reason why TR-FRET assays based around terbium-labeled antibodies or streptavidin perform so well, because there exist a range of donor/acceptor distances, several of which are optimal for measuring FRET.