CD BHK-21 Production Medium
Gibco™
CD BHK-21 Production Medium
Gibco™ CD BHK-21 Production Medium is a chemically defined, protein-free cell culture medium. It is designed to support the growthRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A162770310 kg
A16277011 x 10 L
A16277021 x 100 L
Catalog number A1627703
Price (INR)
281,009.00
Each
Quantity:
10 kg
Price (INR)
281,009.00
Each
Gibco™ CD BHK-21 Production Medium is a chemically defined, protein-free cell culture medium. It is designed to support the growth of BHK-21 cells in serum-free suspension culture. This medium is suitable for large-scale manufacturing applications.
CD BHK-21 Production Medium is manufactured as follows:
WithWithout
• L-glutamine• Phenol Red
• Dry powder formatProtein hydrolysates
Gibco™ CD BHK-21 Production Medium is the first cell culture medium supporting complete serum-free and protein-free processes for BHK-21 cells. The dry powder format is easy to reconstitute, since it has no hydrolysates in the formulation. Adapting cells from serum-containing media is a rapid and simple procedure. BHK-21 cells can achieve high cell densities and maintain high cell viability in this medium.
Gibco™ CD BHK-21 Production Medium is suitable for large-scale suspension culture. Because it does not require supplementation with serum or other undefined components, Gibco™ CD BHK-21 Production Medium eliminates the risks associated with serum, while allowing you to achieve consistent virus titers and simplifying downstream purification.
cGMP Manufacturing
Gibco™ CD BHK-21 Production Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.
Specifications
Cell LineBHK-21
Product TypeProduction Medium
Quantity10 kg
FormPowder
With AdditivesGlutamine, Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).