The GeneArt™ Cryopreservation Kit for Algae is used to preserve algal strains and clones for storage at -80°C for years,Read more
Have Questions?
Catalog Number
Quantity
A24228
100 preps
Catalog number A24228
Price (INR)
-
Quantity:
100 preps
The GeneArt™ Cryopreservation Kit for Algae is used to preserve algal strains and clones for storage at -80°C for years, thus eliminating liquid nitrogen storage and continuous cultures as a way to maintain your clones. Cryopreservation is the optimal choice for preservation and long-term storage of algae, since it minimizes genetic drift, facilitates strain and clone exchange between labs, and helps reduce maintenance labor and costs. Simply grow your cells in the presence of Cryopreservation Reagent A for 2–5 days, harvest, resuspend in Cryopreservation Reagent B, and freeze a small aliquot in one of two recommended freezing containers* at -80°C. The GeneArt™ Cryopreservation Kit for Algae has been used to preserve Chlamydomonas and Chlorella strains with 100% resuscitation after thawing.
• Preserve Chlamydomonas and Chlorella strains and clones for -80°C storage • Eliminate liquid nitrogen storage • Minimize contamination and genetic drift that typically occur in continuous cultures • Ship algal clones on dry ice for exchange between labs
To date, most available methods for algae cell preservation have required liquid nitrogen storage, but this method is inconvenient and expensive. Alternatively, researchers have relied on continuous cultures to maintain algal clones, but this often leads to contamination and genetic drift and is not amenable to shipping. The GeneArt™ Cryopreservation Kit for Algae provides a way to freeze algae cells at -80°C for long-term storage or shipment on dry ice with minimized loss in viability.
*We have used the following freezing containers with this product: • Thermo Scientific™ Mr. Frosty™ Freezing Container • Biocision™ CoolCell™ FTS30 Cell Freezing Container
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Expression SystemAlgae
Product LineGeneArt
Product TypeAlgae Cryopreservation Kit
Quantity100 preps
SpeciesChlamydomonas
Unit SizeEach
Contents & Storage
Contains: • 1 bottle Cryopreservation Reagent A, 5 mL • 1 bottle Cryopreservation Reagent B, 25 mL
Frequently asked questions (FAQs)
What freezing container should I use along with the GeneArt Cryopreservation Kit?
We have used the following freezing containers with this product:
Simply grow your cells in the presence of Cryopreservatin Reagent A for 2-5 days, harvest the cells, and resuspend them in Cryopreservation Reagent B. Follow by freezing the cells at -80 degrees celsius.
What type of algae can the MAX Efficiency Transformation Reagent for Algae be used for?
The MAX Efficiency Transformation Reagent for Algae was developed for transforming the pChlamy_4 vector into the Chlamydomonas reinhardtii 137c cells by electroporation.