RapidFinder™ Haddock ID Kit
Thermo Scientific™

RapidFinder™ Haddock ID Kit

The Thermo Scientific™ RapidFinder™ Haddock ID Kit enables the simple and reliable detection of haddock (Melanogrammus aeglefinus) DNA in food and feed samples.
Have Questions?
Catalog NumberQuantity
A2440548 reactions
Catalog number A24405
Price (INR)
-
Quantity:
48 reactions
The RapidFinder™ Haddock ID Kit enables the simple and reliable detection of Haddock DNA in food and feed samples.

The RapidFinder™ Haddock ID Kit enables the simple and reliable detection of Haddock DNA in food and feed samples. The method works by detection of haddock (Melanogrammus aeglefinus) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Haddock ID Kit detects and provides sensitivity down to 0.01% of haddock (Melanogrammus aeglefinus) DNA. The included positive control in the kit, which comprises 0.1% Haddock DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Haddock DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.
 

Sample preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a complete portfolio of solutions for fish species detection

The RapidFinder™ Haddock ID Kit is just one of our many available kits for the sensitive detection of various fish species in food and feed samples. Other RapidFinder™ Fish species identification kits available include those for the detection of Basa, Gadus, Pollack, Pollock, Tuna, and Specific Tuna.

The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.
For Testing of Food and Environmental Samples Only.
Specifications
DescriptionHaddock ID Kit
For Use With (Equipment)QuantStudio 5™ and 7500 Fast
Product Line

RapidFinder™

Product TypeRapidFinder™
Quantity48 reactions
Sample TypeFood and Environmental Samples
Sensitivity0.01% of Haddock (Melanogrammus aeglefinus) DNA
Shelf Life2 years
SpeciesMelanogrammus aeglefinus
Detection MethodPrimer-probe
For Use With (Application)Food and Beverage
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains 1 tube each of: Haddock Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)