RapidFinder™ Fallow Deer ID Kit
Thermo Scientific™

RapidFinder™ Fallow Deer ID Kit

The Thermo Scientific™ RapidFinder™ Fallow Deer ID Kit enables the simple and reliable detection of fallow deer (Dama dama) DNA in food and feed samples.

Have Questions?
Catalog NumberQuantity
A2441048 reactions
Catalog number A24410
Price (INR)
-
Quantity:
48 reactions

The RapidFinder™ Fallow Deer ID Kit enables the simple and reliable detection of fallow deer (Dama dama) DNA in food and feed samples.

The RapidFinder™ Fallow Deer ID Kit enables the simple and reliable detection of Fallow Deer DNA in food and feed samples. The method works by detection of fallow deer (Dama dama) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Fallow Deer ID Kit detects and provides sensitivity down to 0.01% of Fallow Deer DNA. The included positive control in the kit, which comprises 0.1% Fallow Deer DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Fallow Deer DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Quantification

If a positive result is obtained with the RapidFinder™ Fallow Deer ID Kit, most labs will go on to quantify the amount of Fallow Deer DNA in the sample. This can be accomplished using the RapidFinder™ Fallow Deer ID Kit in conjunction with the RapidFinder™ Quant Multi-Animal Set (Cat. No. A24399).

Part of a complete portfolio of Solutions for Animal Species Detection

The RapidFinder™ Fallow Deer ID Kit is just one of our many available kits for the sensitive detection of various animal species in food and feed samples. Other RapidFinder™ animal species identification kits available include those for the detection of pork, beef, horse, goat, rabbit, water buffalo, duck, sheep, ruminant, chicken, poultry, and turkey.

The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.

For Testing of Food and Environmental Samples Only.
Specifications
DescriptionFallow Deer ID Kit
For Use With (Equipment)QuantStudio 5™ and 7500 Fast
Product LineRapidFinder
Product TypeRapidFinder™
Quantity48 reactions
Sample TypeFood and Environmental Samples
Sensitivity0.01% of Fallow Deer (Dama dama) DNA
Shelf Life2 years
SpeciesDama dama
Detection MethodPrimer-probe
For Use With (Application)Food and Beverage
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains 1 tube each of: Fallow Deer Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)